Skip to Content
Merck
All Photos(2)

Key Documents

O7505

Sigma-Aldrich

Orcein

certified by the Biological Stain Commission, powder

Synonym(s):

Natural Red 28

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

Product Name

Orcein, synthetic, certified by the Biological Stain Commission

biological source

synthetic

grade

certified by the Biological Stain Commission

form

powder

technique(s)

microbe id | staining: suitable

solubility

1 M NaOH: 1 mg/mL

εmax

≥250% at 574-582 nm in NaOH at 0.01 M

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

N(c5c(cc(cc5C)O)O)C1=C(c2[c]([o][c]3c2C(=C(C(=O)C=3)Nc4c(cc(cc4C)O)O)C)=CC1=O)C

InChI

1S/C28H24N2O7/c1-11-5-15(31)7-17(33)25(11)29-27-13(3)23-21(9-19(27)35)37-22-10-20(36)28(14(4)24(22)23)30-26-12(2)6-16(32)8-18(26)34/h5-10,29-34H,1-4H3

InChI key

VPEASJIRGSVXBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Suitability

Certified for use in Darrow′s elastic fiber stain; Orcein method for hepatitis B surface antigen.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kamelija Zarkovic et al.
Redox biology, 4, 109-117 (2015-01-02)
Vascular aging is associated with structural and functional modifications of the arteries, and by an increase in arterial wall thickening in the intima and the media, mainly resulting from structural modifications of the extracellular matrix (ECM) components. Among the factors
Sharma AK and Sharma A
Chromosome Techniques: Theory and Practice (2014)
[Orcein and elastic fibers].
Juan Antonio Barcat
Medicina, 63(5), 453-456 (2003-11-25)
A Fachinelli et al.
Hernia : the journal of hernias and abdominal wall surgery, 15(4), 409-415 (2011-03-15)
The aim of this study was to substantiate the hypothesis as to whether an altered amount of elastic fibers in the linea alba aponeurosis occurs or not in anterior abdominal wall hernias (epigastric, incisional and umbilical), comparing the findings with
C Clementi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(5), 2057-2062 (2008-11-14)
In this paper, three Renaissance tapestries depicting scenes painted by Raffaello Sanzio, conserved at the Vatican Museum, were investigated using in-situ UV-Visible fluorimetric measurements. The results show that this technique is suitable for the detection of natural organic colorants used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service