O5507
Linoleic acid, conjugated
Synonym(s):
CLA, Octadecadienoic acid, conjugated
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352211
NACRES:
NA.77
Recommended Products
impurities
≤1% Linoleic acid
lipid type
omega FAs
storage temp.
−20°C
General description
A mixture of cis- and trans-9,11- and -10,12-octadecadienoic acids. Isomer ratio may vary from lot to lot.
Application
- A targeted nanomodulator capable of manipulating tumor microenvironment against metastasis.: The paper discusses the development of a targeted nanomodulator, including conjugated linoleic acid, to manipulate the tumor microenvironment and inhibit metastasis. The findings suggest significant potential for improved cancer treatments (Tian et al., 2022).
- Hyaluronic Acid-Modified Nanoparticles Self-Assembled from Linoleic Acid-Conjugated Chitosan for the Codelivery of miR34a and Doxorubicin in Resistant Breast Cancer.: The research focuses on hyaluronic acid-modified nanoparticles using linoleic acid-conjugated chitosan for the codelivery of miR34a and doxorubicin. This approach aims to overcome drug resistance in breast cancer treatments (Yang et al., 2022).
Biochem/physiol Actions
Potent inhibitor of breast, skin, and colon carcinogenesis and tumor cell proliferation; inhibits bFGF-induced endothelial cell proliferation; potent antioxidant.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Atique A Behan et al.
Animals : an open access journal from MDPI, 9(7) (2019-07-03)
Rumen protected fats (RPF) are known to improve animal performance without affecting rumen metabolism in sheep. However, comparative effects of prilled fat, prilled fat with lecithin and calcium soap have not been fully studied. Hence this experiment was planned using
Eun-Joung Moon et al.
Oncology reports, 10(3), 617-621 (2003-04-10)
Conjugated linoleic acid (CLA) is a potent inhibitor of mammary carcinogenesis. Cancer cells produce various angiogenic factors which stimulate host vascular endothelial cell mitogenesis and chemotaxis for their growth and metastasis. Basic fibroblast growth factor (bFGF) is a potent angiogenic
Martha A Belury
The Journal of nutrition, 132(10), 2995-2998 (2002-10-09)
Conjugated linoleic acid (CLA) is composed of positional and stereoisomers of octadecadienoate (18:2); it is found in foods derived from ruminants (beef and lamb as well as dairy products from these sources). When a mixture of isomers is fed to
Rui Guo et al.
Frontiers in nutrition, 9, 1026740-1026740 (2022-11-01)
Dietary fat composition is closely associated with the pathological development of alcoholic liver disease (ALD). Fat enriched with saturated fatty acids protects whereas with polyunsaturated fatty acids aggravates alcohol-induced liver injury. However, limited study has addressed how monounsaturated fatty acids
Daeun Lee et al.
Archives of pharmacal research, 43(4), 449-461 (2020-04-03)
Schizophyllan (SPG), produced by Schizophyllum commune, is an exopolysaccharide with multiple academic and commercial uses, including in the food industry and for various medical functions. We previously demonstrated that SPG conjugated with c-Src peptide exerted a significant therapeutic effect on
Articles
Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service