Skip to Content
Merck
All Photos(1)

Key Documents

O111

Sigma-Aldrich

Octoclothepin maleate salt

solid

Synonym(s):

1-(8-Chloro-10,11-dihydrodibenzo[b,f]thiepin-10-yl)-4-methyl-piperazine maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21ClN2S · C4H4O4
CAS Number:
Molecular Weight:
460.97
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

color

white

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: >21 mg/mL

SMILES string

[H]\C(=C(/[H])C(O)=O)C(O)=O.CN1CCN(CC1)C2Cc3ccccc3Sc4ccc(Cl)cc24

InChI

1S/C19H21ClN2S.C4H4O4/c1-21-8-10-22(11-9-21)17-12-14-4-2-3-5-18(14)23-19-7-6-15(20)13-16(17)19;5-3(6)1-2-4(7)8/h2-7,13,17H,8-12H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

GWKBZIADWSOIQV-BTJKTKAUSA-N

Gene Information

Biochem/physiol Actions

D2 dopamine receptor antagonist; 5-HT2 serotonin receptor antagonist.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K P Bøgesø et al.
Journal of medicinal chemistry, 34(7), 2023-2030 (1991-07-01)
Octoclothepin (1) was resolved into its R and S enantiomers via the diastereomeric tartaric acid salts. The enantiomers were shown to be of high optical purity by 1H NMR with use of the chiral shift reagent (R)-(-)-2,2,2-trifluoro-1-(9-anthryl)ethanol. Pharmacological and biochemical
Tetsuji Aoyagi et al.
Shock (Augusta, Ga.), 52(1), 83-91 (2018-07-22)
Excessive inflammation reactions with a cytokine storm in the lungs have historically been thought as the primary cause of fatal acute respiratory distress syndrome (ARDS). However, interruption of inflammatory cytokine activation failed to attenuate ARDS, suggesting that other therapies are
Da Hyun Kim et al.
Journal of dairy science, 102(9), 7707-7716 (2019-07-22)
Maillard reaction products formed from whey protein isolate (WPI) and sugar have been shown to have an anti-inflammatory effect in vitro. Here, we incubated WPI and galactose (GWA) in an aqueous solution at 65°C for 24 h to produce a
Min Chen et al.
Food & function, 10(8), 5080-5090 (2019-07-31)
Acanthopanax trifoliatus (L.) Merr., an edible medicinal plant from Southeast Asia, exerts a wide range of bioactivities, such as anti-inflammatory activity. However, the anti-inflammatory mechanisms of its action and active constituents remain unclear. Herein, the effects of two triterpenoids, namely
Rune Risgaard et al.
Nuclear medicine and biology, 40(1), 135-140 (2012-11-21)
Cerebral α₁-adrenoceptors are a common target for many antipsychotic drugs. Thus, access to positron emission tomography (PET) brain imaging of α₁-adrenoceptors could make important contributions to the understanding of psychotic disorders as well as to the pharmacokinetics and occupancy of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service