Skip to Content
Merck
All Photos(1)

Key Documents

O0391

Sigma-Aldrich

Olopatadine hydrochloride

≥98% (HPLC)

Synonym(s):

(11Z)-11-[3-(Dimethylamino)propylidene]-6,11-dihydrodibenz[b,e]oxepin-2-acetic acid hydrochloride, AL-4943A, KW-4679, Opatanol, Patanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23NO3·HCl
CAS Number:
Molecular Weight:
373.87
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

solubility

H2O: ≥20 mg/mL

storage temp.

room temp

SMILES string

Cl.CN(C)CC\C=C1\c2ccccc2COc3ccc(CC(O)=O)cc13

InChI

1S/C21H23NO3.ClH/c1-22(2)11-5-8-18-17-7-4-3-6-16(17)14-25-20-10-9-15(12-19(18)20)13-21(23)24;/h3-4,6-10,12H,5,11,13-14H2,1-2H3,(H,23,24);1H/b18-8-;

InChI key

HVRLZEKDTUEKQH-NOILCQHBSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Olopatadine is a dual acting histamine H1-receptor antagonist and mast cell stabilizer.
Olopatadine is a histamine H1 receptor antagonist and mast cell stabilizer. The low level of occupancy H1 receptors in the brain explains the low sedation effect of olopatidine. It is believed that olopatidine is a substrate for P-glycoprotein, which limits its brain penetration.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eli O Meltzer et al.
Allergy and asthma proceedings, 32(3), 213-220 (2011-04-12)
Allergic rhinitis (AR) is highly prevalent in children. Olopatadine, 0.6% nasal spray (olopatadine) is approved for the relief of seasonal allergic rhinitis (SAR) symptoms in children 6 years of age and older. The objective of this study is to provide
Fuad M Baroody et al.
American journal of rhinology & allergy, 27(1), 48-53 (2013-02-15)
Nasal allergen challenge (NAC) leads to a nasal ocular reflex, which is augmented by allergic inflammation. This study was designed to confirm our previous observation that an intranasal steroid inhibits the nasal ocular reflex and to show that histamine does
Yukari Okubo et al.
The Journal of dermatological treatment, 24(2), 153-160 (2011-08-04)
An increased dose of the prescribed drug or a change of the drug is recommended in the treatment for the patients with urticaria refractory to the standard dose of antihistamines. Efficacy and safety of doubling the dose of cetirizine were
Masahiko Shimura et al.
American journal of ophthalmology, 151(4), 697-702 (2011-01-25)
To evaluate the effectiveness of pre-seasonal treatment with topical olopatadine on the reduction of clinical symptoms of seasonal allergic conjunctivitis (SAC). Prospective interventional case series. Eleven patients with SAC received topical olopatadine in one eye at least two weeks before
Peter K Smith et al.
American journal of rhinology & allergy, 25(4), e149-e152 (2011-08-09)
Vasomotor rhinitis (VMR) is a hypersensitivity syndrome with heightened reactivity to environmental triggers. Twenty-two patients with severe VMR were treated nasally with either normal saline or 0.6% olopatadine and challenged nasally with a hyperosmolar mannitol solution. Treatment with 0.6% olopatadine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service