Skip to Content
Merck
All Photos(1)

Key Documents

N8768

Sigma-Aldrich

Natriuretic Peptide, C-Type

≥95% (HPLC)

Synonym(s):

CNP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C93H157N27O28S3
CAS Number:
Molecular Weight:
2197.60
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

composition

Peptide content, ≥65%

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@@H]1NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CC(C)C)NC(=O)CNC(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](CSSC[C@H](NC(=O)CNC(=O)[C@H](CC(C)C)NC(=O)CNC(=O)[C@H](CO)NC(=O)[C@H](CCSC)NC(=O)[C@H](CO)NC(=O)CNC1=O)C(O)=O)NC(=O)CNC(=O)[C@H](CCCCN)NC(=O)[C@H](CO)NC(=O)[C@H](CC(C)C)NC(=O)CN

InChI

1S/C93H157N27O28S3/c1-12-52(10)76-91(146)104-42-72(127)108-65(44-122)88(143)114-57(26-30-149-11)82(137)118-64(43-121)80(135)103-39-70(125)106-58(31-48(2)3)78(133)101-41-74(129)110-68(92(147)148)47-151-150-46-67(109-73(128)40-100-77(132)54(23-16-18-27-94)111-89(144)66(45-123)119-85(140)59(32-49(4)5)105-69(124)37-96)90(145)116-62(35-53-21-14-13-15-22-53)79(134)102-38-71(126)107-60(33-50(6)7)84(139)112-55(24-17-19-28-95)81(136)115-61(34-51(8)9)86(141)117-63(36-75(130)131)87(142)113-56(83(138)120-76)25-20-29-99-93(97)98/h13-15,21-22,48-52,54-68,76,121-123H,12,16-20,23-47,94-96H2,1-11H3,(H,100,132)(H,101,133)(H,102,134)(H,103,135)(H,104,146)(H,105,124)(H,106,125)(H,107,126)(H,108,127)(H,109,128)(H,110,129)(H,111,144)(H,112,139)(H,113,142)(H,114,143)(H,115,136)(H,116,145)(H,117,141)(H,118,137)(H,119,140)(H,120,138)(H,130,131)(H,147,148)(H4,97,98,99)/t52-,54-,55-,56-,57-,58-,59-,60-,61-,62-,63-,64-,65-,66-,67-,68-,76-/m0/s1

InChI key

UHSBKBYWHCBDCJ-DYGWFTAGSA-N

Gene Information

human ... NPPC(4880)

Amino Acid Sequence

Gly-Leu-Ser-Lys-Gly-Cys-Phe-Gly-Leu-Lys-Leu-Asp-Arg-Ile-Gly-Ser-Met-Ser-Gly-Leu-Gly-Cys [Disulfide Bridge: 6-22]

General description

Natriuretic Peptide, C-Type (NPPC/ CNP) is a 22 amino acid peptide encoded by NPPC gene mapped to human chromosome 2q37.1. It belongs to the natriuretic peptide family.

Biochem/physiol Actions

C-type natriuretic peptide is produced in the hypothalamus, anterior pituitary, and most major endocrine glands. It has potent venodilatory and coronary vasodilatory effects, but minimal effects on renal function.
Natriuretic Peptide, C-Type (NPC) plays a vital role in regulation of blood pressure and body fluid homeostasis. NPC-mediated signaling is essential for endochondral ossification. NPC acts as a ligand for natriuretic peptide receptor B (NPR-B). NPC interacts with receptors on vascular smooth muscle cells (VSMCs) and functions as an autocrine/paracrine endothelium-derived hormone. It is also implicated in the endothelial dysfunction and regulation of adipogenesis, which has a crucial role in the development of obesity in childhood. NPC has potential as a marker for obesity and bone injury induced by steroid hormones. In vitro studies show that NPC is involved in the inhibition of smooth muscle cell proliferation, natriuresis, diuresis, vasodilation and inhibition of the renin- angiotensin system. NPC is associated with the development of myocardial infarction and heart failure. Plasma NPC levels can be used as a biomarker for the therapeutic efficacy of metoprolol on postural tachycardia syndrome (POTS) in children.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyung-Bon Lee et al.
Biology of reproduction, 88(2), 42-42 (2012-12-21)
Natriuretic peptide type C (NPPC) and its receptor natriuretic peptide receptor 2 (NPR2) regulate cGMP in ovarian follicles and participate in maintaining oocyte meiotic arrest. We investigated the regulation of Nppc expression in mouse granulosa cells in vivo and in
Fanny Caputo et al.
Scientific reports, 7(1), 4636-4636 (2017-07-07)
Cerium oxide nanoparticles (CNPs) are novel synthetic antioxidant agents proposed for treating oxidative stress-related diseases. The synthesis of high-quality CNPs for biomedical applications remains a challenging task. A major concern for a safe use of CNPs as pharmacological agents is
Y Takei
International review of cytology, 194, 1-66 (1999-09-24)
The natriuretic peptide (NP) system consists of three types of hormones [atrial NP (ANP), brain or B-type NP (BNP), and C-type NP (CNP)] and three types of receptors [NP receptor (R)-A, NPR-B, and NPR-C]. ANP and BNP are circulating hormones
C-type natriuretic peptide is closely associated to obesity in Caucasian adolescents.
Del Ry S
Clinica Chimica Acta; International Journal of Clinical Chemistry, 460, 172-177 (2016)
C-type natriuretic peptide: the endothelial component of the natriuretic peptide system.
Chen HH and Burnett JC Jr
Journal of Cardiovascular Pharmacology, 32 Suppl 3, S22-S28 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service