Skip to Content
Merck
All Photos(1)

Key Documents

N8523

Sigma-Aldrich

Monoclonal Anti-NF-κB antibody produced in mouse

clone NF-12, ascites fluid

Synonym(s):

Anti-CVID12, Anti-EBP-1, Anti-KBF1, Anti-NF-kB, Anti-NF-kB1, Anti-NF-kappa-B1, Anti-NF-kappaB, Anti-NF-kappabeta, Anti-NFKB-p105, Anti-NFKB-p50, Anti-NFkappaB

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

NF-12, monoclonal

mol wt

antigen 65 kDa

contains

15 mM sodium azide

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable
microarray: suitable
western blot: 1:1,000 using a whole cell extract from a mouse fibroblast cell line

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NFKB1(4790)
mouse ... Nfkb1(18033)

General description

Monoclonal Anti-NF-κB p65 (mouse IgG1 isotype) is derived from the NF-12 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from a BALB/c mouse immunized with a recombinant, mouse NF-κB p65 fragment. Nuclear factor κB (NF-κB1) is a member of NF-κB/Rel family of transcription factors.
Nuclear factor of kappa light polypeptide gene enhancer in B cells 1, p105 (NFKB1) is a subunit of nuclear factor κB (NF-κB) and exists as a dimer. It is expressed in the cytosol.

Immunogen

recombinant mouse NF-κB p65 fragment (C-terminal, 151 amino acids).

Application

Monoclonal Anti-NF-κB antibody produced in mouse has been used in immunohistochemistry and Western blotting.

Biochem/physiol Actions

Nuclear factor kappa-B (NF-κB1) has an ability to affect transcription of κB reporter genes positively or negatively when assayed following transfection.
Nuclear factor of kappa light polypeptide gene enhancer in B cells 1, p105 (NFKB1) has a role in hippocampal neurogenesis. It regulates the signaling by nuclear factor κB (NF-κB). Deletion of the gene in mice leads to defects in innate and adaptive immunity. The protein acts as a mediator of apoptotic response to S-phase DNA damage and replication stress. NFKB1 modulates the structural plasticity of cortical neurons. Knockout of the protein in mice has been shown to cause liver apoptosis and embryonic death.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deletion of nuclear factor-?B p50 upregulates fatty acid utilization and contributes to an anti-obesity and high-endurance phenotype in mice.
Minegishi Y
American Journal of Physiology. Endocrinology and Metabolism, 309(6), E523-E533 (2015)
Loss of Nfkb1 leads to early onset aging
Giovanna M. Bernal
Aging, 6(11), 931?942-931?942 (2014)
NF-kappaB family of transcription factors: central regulators of innate and adaptive immune functions
Caamano J and Hunter C A
Clinical Microbiology Reviews, 15(3), 414-429 (2002)
A Combination of Podophyllotoxin and Rutin Attenuates Radiation Induced Gastrointestinal Injury by Negatively Regulating NF-?B/p53 Signaling in Lethally Irradiated Mice
Bhargab Kalita
PLoS ONE, 11(12), e0168525-e0168525 (2016)
Cell autonomous and noncell-autonomous role of NF-?B p50 in astrocyte-mediated fate specification of adult neural progenitor cells.
Cvijetic S
Glia, 65, 169-181 (2017)

Articles

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service