Skip to Content
Merck
All Photos(1)

Key Documents

N5504

Sigma-Aldrich

Naphazoline hydrochloride

Synonym(s):

2-(1-Naphthylmethyl)imidazoline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N2 · HCl
CAS Number:
Molecular Weight:
246.74
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

originator

Allergan

SMILES string

Cl.C1CN=C(Cc2cccc3ccccc23)N1

InChI

1S/C14H14N2.ClH/c1-2-7-13-11(4-1)5-3-6-12(13)10-14-15-8-9-16-14;/h1-7H,8-10H2,(H,15,16);1H

InChI key

DJDFFEBSKJCGHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

α-adrenoceptor agonist; imidazoline receptor agonist; vasoconstrictor.
Naphazoline hydrochloride is a sympathomimetic agent. It is also considered as a conjunctival decongestant.

Features and Benefits

This compound was developed by Allergan. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M Gelardi et al.
Journal of biological regulators and homeostatic agents, 23(2), 79-84 (2009-07-11)
A wide variety of nasal irrigation systems are currently available for improving nasal symptoms, but few studies have compared their effectiveness with respect to patient age and type of nasal disease. This pilot study aims to compare the efficacy of
Marie Granier et al.
Canadian journal of anaesthesia = Journal canadien d'anesthesie, 56(2), 102-108 (2009-02-28)
Septorhinoplasty is a traumatic procedure that is associated with epistaxis and postoperative pain. The primary objective of this randomized double-blind controlled trial was to determine whether intranasal 5% lidocaine plus naphazoline decreases postoperative pain and lessens the use of rescue
Nan-Nan Wang et al.
Luminescence : the journal of biological and chemical luminescence, 24(3), 178-182 (2009-03-03)
A sensitive and simple flow-injection chemiluminescence (FI-CL) method, which was based on the CL intensity generated from the redoxreaction of potassium permanganate (KMnO4)-formaldehyde in vitriol (H2SO4) medium, has been developed, validated and applied for the determination of naphazoline hydrochloride and
Yuriko Nagane et al.
Muscle & nerve, 44(1), 41-44 (2011-04-15)
When treating ocular myasthenia gravis (MG), the risk/benefit profile of corticosteroids is unclear, and acetylcholinesterase inhibitors are not very effective. We examined the efficacy of topical naphazoline in the treatment of myasthenic blepharoptosis. Sixty MG patients with blepharoptosis (32 with
Eman Y Z Frag et al.
The Analyst, 136(2), 332-339 (2010-10-21)
This paper describes the development of screen-printed (SPE) and carbon paste (CPE) sensors for the rapid and sensitive quantification of naphazoline hydrochloride (NPZ) in pharmaceutical formulations. This work compares the electroactivity of conventional carbon paste and screen-printed carbon paste electrodes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service