Skip to Content
Merck
All Photos(1)

Key Documents

N4143

Sigma-Aldrich

Neurokinin B

≥95% (HPLC)

Synonym(s):

β-Neurokinin, Neuromedin K

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H79N13O14S2
CAS Number:
Molecular Weight:
1210.42
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥95% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(CC(C)C)NC(=O)CNC(=O)C(NC(=O)C(Cc1ccccc1)NC(=O)C(Cc2ccccc2)NC(=O)C(CC(O)=O)NC(=O)C(Cc3cnc[nH]3)NC(=O)C(CCSC)NC(=O)C(N)CC(O)=O)C(C)C)C(N)=O

InChI

1S/C55H79N13O14S2/c1-30(2)21-38(50(77)62-36(47(57)74)17-19-83-5)61-43(69)28-59-55(82)46(31(3)4)68-54(81)40(23-33-15-11-8-12-16-33)65-51(78)39(22-32-13-9-7-10-14-32)64-53(80)42(26-45(72)73)67-52(79)41(24-34-27-58-29-60-34)66-49(76)37(18-20-84-6)63-48(75)35(56)25-44(70)71/h7-16,27,29-31,35-42,46H,17-26,28,56H2,1-6H3,(H2,57,74)(H,58,60)(H,59,82)(H,61,69)(H,62,77)(H,63,75)(H,64,80)(H,65,78)(H,66,76)(H,67,79)(H,68,81)(H,70,71)(H,72,73)

InChI key

NHXYSAFTNPANFK-UHFFFAOYSA-N

Gene Information

human ... TAC3(6866)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Asp-Met-His-Asp-Phe-Phe-Val-Gly-Leu-Met-NH2

Biochem/physiol Actions

Neurokinin B is a ten-residue peptide. It is an endogenous tachykinin agonist with preference for the NK-3 receptor. It is a potent bronchoconstrictor and may have a neuromodulatory role in the brain. Neurokinin B has a role in slow synaptic or non-synaptic communication.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neurokinin B- and specific tachykinin NK3 receptor agonists-induced airway hyperresponsiveness in the guinea-pig
Samira D
British Journal of Pharmacology (2000)
The Tachykinin Peptide Neurokinin B Binds Copper Forming an Unusual [CuII(NKB)2] Complex and Inhibits Copper Uptake into 1321N1 Astrocytoma Cells
ACS Chemical Neuroscience (2013)
Neurokinin B Acts via the Neurokinin-3 Receptor in the Retrochiasmatic Area to Stimulate Luteinizing Hormone Secretion in Sheep
Endocrinology (2010)
Michael N Lehman et al.
Endocrinology, 151(8), 3479-3489 (2010-05-27)
Recently, a subset of neurons was identified in the arcuate nucleus of the hypothalamus that colocalize three neuropeptides, kisspeptin, neurokinin B, and dynorphin, each of which has been shown to play a critical role in the central control of reproduction.
F Ruiz-Pino et al.
Endocrinology, 153(10), 4818-4829 (2012-07-24)
Neurokinin B (NKB), encoded by Tac2 in rodents, and its receptor, NK3R, have recently emerged as important regulators of reproduction; NKB has been proposed to stimulate kisspeptin output onto GnRH neurons. Accordingly, NKB has been shown to induce gonadotropin release

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service