Skip to Content
Merck
All Photos(1)

Documents

MAK135

Sigma-Aldrich

ADP/ATP Ratio Assay Kit

sufficient for 100 tests (bioluminescent)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 tests (bioluminescent)

application(s)

pharmaceutical

detection method

chemiluminescent

relevant disease(s)

cancer

storage temp.

−20°C

General description

Changes in the ADP/ATP ratio have been used to differentiate modes of cell death and viability. Increased levels of ATP and decreased levels of ADP signify proliferating cells. Conversely, decreased levels of ATP and increased levels of ADP represent apoptotic or necrotic cells where the decrease in ATP and increase in ADP are much more pronounced in necrosis versus apoptosis.

Application


  • LOC554202 contributes to chordoma progression by sponging miR-377-3p and up-regulating SMAD3.: This article investigates the molecular mechanisms by which LOC554202 facilitates chordoma progression through miR-377-3p and SMAD3 regulation. The ADP/ATP Ratio Assay Kit was utilized to assess the metabolic impact of these molecular changes on cell viability and proliferation (Xu et al., 2023).

  • FOXG1 improves mitochondrial function and promotes the progression of nasopharyngeal carcinoma.: This study explores how FOXG1 enhances mitochondrial function, thereby promoting nasopharyngeal carcinoma progression. The ADP/ATP Ratio Assay Kit was employed to measure mitochondrial function and energy metabolism changes, providing insights into the bioenergetic profile of cancer cells (Xi et al., 2021).

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the detection of apoptosis and necrosis in cells and for the studying the effects of compounds on cellular proliferation.

Principle

The ADP/ATP Ratio Assay kit provides a simple and direct procedure for measuring ADP and ATP levels in cells for the screening of apoptosis, necrosis, and cell proliferation. The assay involves two steps. In the first step, the working reagent lyses cells to release ATP and ADP. In the presence of luciferase, ATP immediately reacts with the Substrate D-luciferin to produce light. The light intensity is a direct measure of the intracellular ATP concentration.

Luciferase
ATP + D-Luciferin + O2 ----------> oxyluciferin + AMP + PPi + CO2 + light

In the second step, the ADP is converted to ATP through an enzyme reaction. This newly formed ATP then reacts with the D-luciferin as in the first step. The second light intensity measured represents the total ADP and ATP concentration in the sample.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nidal Zeineh et al.
Cells, 8(7) (2019-07-13)
The 18 kDa translocator protein (TSPO) is an initiator of the mitochondrial apoptosis cascade. Cigarette smoke (CS) exposure provokes alterations in TSPO expression as well as upregulation of its related functions such as mitochondrial membrane potential (ΔψM) and reactive oxygen
Sangiliyandi Gurunathan et al.
Nanomaterials (Basel, Switzerland), 8(6) (2018-06-06)
The unique properties of gold nanoparticles (AuNPs) have attracted much interest for a range of applications, including biomedical applications in the cosmetic industry. The current study assessed the anti-oxidative effect of AuNPs against retinoic acid (RA)-induced loss of cell viability;
Aline G Cozer et al.
Lipids, 51(11), 1303-1307 (2016-10-25)
The present work assesses in vitro the role of human Stanniocalcin 1 (hSTC-1) in glucose metabolism in white retroperitoneal adipose tissue (WRAT) from fed rat. In the fed state, hSTC1 increases the incorporation of
Claudia Lin-Kar Hung et al.
Molecular biology of the cell, 29(23), 2809-2820 (2018-09-27)
The huntingtin protein participates in several cellular processes that are disrupted when the polyglutamine tract is expanded beyond a threshold of 37 CAG DNA repeats in Huntington's disease (HD). Cellular biology approaches to understand these functional disruptions in HD have
Sangiliyandi Gurunathan et al.
Nanomaterials (Basel, Switzerland), 9(7) (2019-07-05)
Graphene, a two-dimensional carbon sheet with single-atom thickness, shows immense promise in several nanoscientific and nanotechnological applications, including in sensors, catalysis, and biomedicine. Although several studies have shown the cytotoxicity of graphene oxide in different cell types, there are no

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service