Skip to Content
Merck
All Photos(3)

Documents

MAK093

Sigma-Aldrich

Phosphofructokinase (PFK) Activity Colorimetric Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

PFK Activity Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric tests

detection method

colorimetric

relevant disease(s)

cancer; orthopedic diseases; aging/geriatric diseases

storage temp.

−20°C

General description

Phosphofructokinase (PFK) catalyzes the third step of glycolysis, the conversion of fructose-6-phosphate to fructose-1,6-diphosphate, the rate limiting step of glycolysis. PFK activity is highly regulated by multiple cofactors in addition to post-translational modifications and can be used to measure glycolytic flux in tissues. Deficiencies in PFK activity can result in the glycogen storage disease, glycogenosis type VII (Tarui′s disease), which is a glycogen storage disease.

Application

Phosphofructokinase (PFK) Activity Colorimetric Assay Kit has been used to measure the activity of phosphofructokinase.

Suitability

Suitable for the measurement of phosphofructokinase activity, analysis of glucose metabolism and cell signaling and screening anti-cancer drugs in animal tissues (liver, brain, heart, muscles etc.) and cell culture (adherent or suspension cells).

Principle

The Phosphofructokinase Colorimetric Assay kit provides a simple and direct procedure for measuring PFK activity in a variety of samples. PFK activity is determined by a coupled enzyme assay, in which fructose-6-phosphate and ATP is converted to fructose-1,6-diphosphate and ADP by PFK. The ADP is converted by the enzyme mix to AMP and NADH. The resulting NADH reduces a colorless probe resulting in a colorimetric (450 nm) product proportional to the PFK activity present. One unit of PFK is the amount of enzyme that will generate 1.0 μmole of NADH per minute at pH 7.4 at 37 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neng Tang et al.
Biochemical and biophysical research communications, 526(1), 1-7 (2020-03-21)
The mechanisms underlying the initiation and proliferation of liver regeneration (LR) has been extensively studied using the partial hepatectomy (PHx) model, while little is known about the termination of LR. PP2Acα (protein phosphatase 2 A catalytic subunit α isoform) is
Kai Zhou et al.
Cell death & disease, 9(10), 988-988 (2018-09-27)
Plastic phenotype convention between glioma stem cells (GSCs) and non-stem tumor cells (NSTCs) significantly fuels glioblastoma heterogeneity that causes therapeutic failure. Recent progressions indicate that glucose metabolic reprogramming could drive cell fates. However, the metabolic pattern of GSCs and NSTCs
Manipulation of a glycolytic regulator alters growth and carbon partitioning in the marine diatom Thalassiosira pseudonana.
Abbriano R, et al.
Algal research, 32, 250-258 (2018)
Emilio Bueno et al.
Nature microbiology, 3(12), 1346-1353 (2018-10-03)
To survive and proliferate in the absence of oxygen, many enteric pathogens can undergo anaerobic respiration within the host by using nitrate (NO3-) as an electron acceptor1,2. In these bacteria, NO3- is typically reduced by a nitrate reductase to nitrite
Endurance exercise causes adverse changes in some hematological and physio-biochemical indices in ponies under high altitude stress condition.
Vivek P, et al.
The Indian journal of animal sciences., 88(2), 222-228 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service