Skip to Content
Merck
All Photos(2)

Key Documents

MAK067

Sigma-Aldrich

Malate Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric tests

detection method

colorimetric

storage temp.

−20°C

General description

L-Malate is a tricarboxylic acid cycle intermediate and a critical component of the malate-aspartate shuttle. The Malate-aspartate shuttle is required for transporting reducing equivalents across the mitochondrial membrane for use in oxidative phosphorylation. In lower organisms, malate is converted to lactate during malolactic fermentation with the formation of CO2. Malate is frequently used as an additive in the food and pharmaceutical industries, so quantitating malate is important in the manufacturing of beer, wine, cheese, and fruits. The Malate Assay kit is an easy and sensitive assay to measure the malate level in a variety of samples.

In this kit, malate is oxidized to generate a product specifically detected by colorimetric (450 nm) assay. Typical sensitivities of detection for this kit are 1-35 nmol of malate.

Application

Malate Assay Kit has been used to determine the concentration of malate in samples.

Suitability

Suitable for the measurement of L(-) Malate level in a variety of samples including tissue, cells and food and beverages (for example beer, wine, cheese and fruit)

Principle

Malate concentration is determined by an enzymatic assay, which results in a colorimetric (450 nm) product, proportional to the malate present. Typical detection range for this kit is 5–25 nmoles of malate.

replaced by

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen R Thom et al.
Free radical biology & medicine, 106, 406-416 (2017-03-16)
We hypothesized that elevations of carbon dioxide (CO2) commonly found in modern buildings will stimulate leukocytes to produce microparticles (MPs) and activate the nucleotide-binding domain-like receptor 3 (NLRP3) inflammasome due to mitochondrial oxidative stress. Human and murine neutrophils generate MPs
Melatonin alleviates adipose inflammation through elevating α-ketoglutarate and diverting adipose-derived exosomes to macrophages in mice.
Liu Z, et al.
Journal of Pineal Research, 64(1), e12455-e12455 (2018)
Lingjie Sang et al.
Nature metabolism, 3(1), 90-106 (2021-01-06)
Organelles use specialized molecules to regulate their essential cellular processes. However, systematically elucidating the subcellular distribution and function of molecules such as long non-coding RNAs (lncRNAs) in cellular homeostasis and diseases has not been fully achieved. Here, we reveal the
Nanmu Yang et al.
Journal of cellular physiology, 236(5), 3863-3880 (2020-12-12)
Cancer cells sustain high levels of glycolysis and glutaminolysis via reprogramming of intracellular metabolism, which represents a driver of hepatocellular carcinoma (HCC) progression. Understanding the mechanisms of cell metabolic reprogramming may present a new basis for liver cancer treatment. Herein
Michael W Shane et al.
Plant physiology, 161(4), 1634-1644 (2013-02-15)
Accumulating evidence indicates important functions for phosphoenolpyruvate (PEP) carboxylase (PEPC) in inorganic phosphate (Pi)-starved plants. This includes controlling the production of organic acid anions (malate, citrate) that are excreted in copious amounts by proteoid roots of nonmycorrhizal species such as

Articles

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service