Skip to Content
Merck
All Photos(4)

Key Documents

M9376

Sigma-Aldrich

Methyl α-D-glucopyranoside

≥99% (GC)

Synonym(s):

Methyl α-D-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O6
CAS Number:
Molecular Weight:
194.18
Beilstein:
81568
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

≥99% (GC)

form

solid

optical activity

[α]20/D 156 to 160 °, c = 10% (w/v) in water

technique(s)

gas chromatography (GC): suitable

color

white

solubility

water: 50 mg/mL, clear, colorless

storage temp.

room temp

SMILES string

CO[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C7H14O6/c1-12-7-6(11)5(10)4(9)3(2-8)13-7/h3-11H,2H2,1H3/t3-,4-,5+,6-,7+/m1/s1

InChI key

HOVAGTYPODGVJG-ZFYZTMLRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Different concentrations of methyl α-D-glucopyranoside were used to vary echo decay times in a study that assessed the effects of cryoprotection on the structure and activity of p21ras. Methyl α-D-glucopyranoside has also been used in a study to investigate saccharide-mediated protection of chaotropic-induced deactivation of concanavalin A.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

D N Figlas et al.
Archives of biochemistry and biophysics, 340(1), 154-158 (1997-04-01)
This work provides evidence of a physical instance in which some proteins that are usually inactivated under strong chaotropic conditions may become fully resistant through the occupancy of their binding sites with suitable ligands. In this regard, we found that
C J Halkides et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 134(1), 142-153 (1998-09-19)
Electron spin-echo envelope modulation (ESEEM) spectroscopy is widely used to investigate the active sites of biological molecules in frozen solutions. Various cryoprotection techniques, particularly the addition of co-solvents, are commonly employed in the preparation of such samples. In conjunction with
Mikkel G Jørgensen et al.
Journal of bacteriology, 191(4), 1191-1199 (2008-12-09)
Toxin-antitoxin (TA) loci are common in free-living bacteria and archaea. TA loci encode a stable toxin that is neutralized by a metabolically unstable antitoxin. The antitoxin can be either a protein or an antisense RNA. So far, six different TA
Yu Jin Lee et al.
American journal of physiology. Renal physiology, 296(6), F1405-F1416 (2009-02-13)
Recent studies demonstrated that endoplasmic reticulum (ER) stress regulates glucose homeostasis and that ER stress preconditioning which induces an adaptive, protective unfolded protein response (UPR) offers cytoprotection against nephrotoxins. Thus the aim of the present study was to use renal
Carmen Castaneda-Sceppa et al.
Journal of physiology and biochemistry, 66(2), 105-115 (2010-04-30)
The present study explored whether calcitriol plays a role in the regulation of sodium-dependent glucose transporter protein 1 (SGLT1) activity. For this purpose, alpha-methyl glucoside (AMG) uptake in stable transfected Chinese hamster ovary (CHO-G6D3) cells expressing rabbit SGLT1 (rbSGLT1) was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service