Skip to Content
Merck
All Photos(2)

Key Documents

M8527

Sigma-Aldrich

4-Methylumbelliferyl α-L-fucopyranoside

fluorogenic, ≥97% (TLC), powder

Synonym(s):

4-Methylumbelliferyl alpha-L-fucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18O7
CAS Number:
Molecular Weight:
322.31
Beilstein:
8160514
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.21

Product Name

4-Methylumbelliferyl α-L-fucopyranoside, ≥97% (TLC)

Quality Level

Assay

≥97% (TLC)

form

powder

solubility

DMF: 100 mg/mL, clear, colorless to faintly yellow

fluorescence

λex 360 nm; λem 449 nm (Reaction product)

storage temp.

−20°C

SMILES string

C[C@@H]1O[C@@H](Oc2ccc3C(C)=CC(=O)Oc3c2)[C@@H](O)[C@H](O)[C@@H]1O

InChI

1S/C16H18O7/c1-7-5-12(17)23-11-6-9(3-4-10(7)11)22-16-15(20)14(19)13(18)8(2)21-16/h3-6,8,13-16,18-20H,1-2H3/t8-,13+,14+,15-,16-/m0/s1

InChI key

CQKHENXHLAUMBH-CRLRYRHBSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

4-Methylumbelliferyl α-L-fucopyranoside has been used:
  • as a substrate in α-L-fucosidase 1 (α-L-FUCA-1) enzyme assays to convert it to the fluorescent product 4-methylumbelliferone (4MU) for finding the distribution of the membrane-associated α-L-FUCA-1 of human sperm
  • as a fluorogenic substrate in enzyme assays to evaluate the stability of both the seminal plasma and membrane-associated α-l-fucosidase
  • as a substrate to measure the enzymatic activity of α-L-FUCA-1 of cell extracts from TPC-1, BCPAP, 8505C, and CAL62 cell lines

Substrates

Substrate for α-L-fucosidase

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Giuliani et al.
Glycoconjugate journal, 10(6), 447-452 (1993-12-01)
The behaviour of a highly purified alpha-L-fucosidase (E.C. 3.2.1.51) extracted from octopus hepatopancreas was studied with phospholipid vesicles composed of phosphatidylcholine (PC) and phosphatidylserine (PS) containing the fucosylated ganglioside FucGM1, a potential natural substrate of the enzyme. The substrate recognition
Jennifer J Venditti et al.
Molecular reproduction and development, 74(6), 758-766 (2006-11-30)
Two distinctive isoforms of the enzyme alpha-L-fucosidase are found within human semen in substantial amounts, suggesting specialized functions during reproduction. The membrane-associated isozyme of human sperm cells was previously characterized biochemically, and here we report on its subcellular localization. Intact
Giancarlo Vecchio et al.
Oncotarget, 8(16), 27075-27092 (2017-04-14)
Glycans containing α-L-fucose participate in diverse interactions between cells and extracellular matrix. High glycan expression on cell surface is often associated with neoplastic progression. The lysosomal exoenzyme, α-L-fucosidase-1 (FUCA-1) removes fucose residues from glycans. The FUCA-1 gene is down-regulated in
M Masserini et al.
The Biochemical journal, 229(3), 595-603 (1985-08-01)
alpha-L-Fucosidase, prepared in highly purified form (Mr 70 000-74 000) from Octopus hepatopancreas, was able to hydrolyse a fucose-containing ganglioside, namely Fuc-GM1 (II3NeuAc,IV2Fuc-GgOse4-Cer). The enzyme showed an irregular kinetic behaviour (v/[S] and v/[E] relationships following sigmoidal curves) when working on
Jennifer J Venditti et al.
International journal of andrology, 32(5), 556-562 (2008-06-05)
Previous reports from this laboratory documented the existence of two novel isoforms of alpha-L-fucosidase in human semen and showed that membrane-associated alpha-L-fucosidase is cryptically held within the acrosomal compartment and enriched within the sperm equatorial segment. The occurrence of these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service