Skip to Content
Merck
All Photos(1)

Key Documents

M7769

Sigma-Aldrich

IgG2a, Kappa from murine myeloma

clone UPC 10, ascites fluid, lyophilized powder

Synonym(s):

Mouse IgG2a-κ

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

clone

UPC 10, monoclonal

form

lyophilized powder

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The UPC-10 tumor line that produces mouse IgG2a, K is a tetramethylpentadecane (pristane) induced plasmacytoma originated and carried intraperitoneally in BALB/c mice. The hapten binding specificity of the UPC-10 line is for β-2-6-linked fructosan. The ascites produced from the UPC-10 tumor line contains IgG2a, K in addition to normal levels of other mouse immunoglobulins and serum proteins. The ascites fluid is clarified by centrifugation and filtration. IgG2a (immunoglobulin G 2a) is one of the subclasses of rat immunoglobulins, which also include IgG1, IgG2b and IgG2c.

Specificity

Antigen Specificity: β-2,6-fructosan

Trace amounts of lambda chain may be present.

Immunogen

The UPC-10 tumor line that produces mouse IgG2a, K is a tetramethylpentadecane (pristane) induced plasmacytoma originated and carried intraperitoneally in BALB/c mice.

Application

IgG2a, Kappa from murine myeloma has been used in immunofluorescence and immunohistochemistry.

Biochem/physiol Actions

The IgG2a (immunoglobulin G 2a) plays a vital role in stimulating autoimmunity in mice. This gene is expressed at high level during virus infection. IgG2a offers host defence through activation of complement system. This antibody has specificity towards Fc receptors expressed on phagocytes and stimulates antibody-dependent cell mediated cytotoxicity. IgG2a also induces macrophage-mediated opsonophagocytosis, which has been observed in response to influenza virus and leads to its clearance from the infected cells.

Physical form

The product provided is lyophilized from 0.01 M phosphate buffered saline, pH 7.2, with no preservatives added.

Preparation Note

Storage and Stability: Store the product at 2-8 °C. After reconstitution, the solution may be stored frozen in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teizo Asano et al.
Antibodies (Basel, Switzerland), 11(4) (2022-12-23)
The epithelial cell adhesion molecule (EpCAM) is a stem cell and carcinoma antigen, which mediates cellular adhesion and proliferative signaling by the proteolytic cleavage. In contrast to low expression in normal epithelium, EpCAM is frequently overexpressed in various carcinomas, which
N Reddy et al.
Cancer immunology, immunotherapy : CII, 46(1), 25-33 (1998-03-31)
We reported previously that the blood clearance of injected mouse IgG2a was extremely rapid in many strains of nude and nu/+ mice. In an attempt to determine the cause of this phenomenon, the levels of endogenous IgG2a in the blood
S Escotte et al.
The European respiratory journal, 21(4), 574-581 (2003-05-24)
Inhaled fluticasone propionate (FP) is widely used to reduce pulmonary inflammation in chronic obstructive pulmonary disease, but the potential effects of FP on airway epithelial cells from patients with cystic fibrosis (CF) are unknown. In CF disease, a nonregulated inflammatory
Juan Feng et al.
eLife, 10 (2021-12-24)
HAP2 is a transmembrane gamete fusogen found in multiple eukaryotic kingdoms and is structurally homologous to viral class II fusogens. Studies in Plasmodium have suggested that HAP2 is an attractive target for vaccines that block transmission of malaria. HAP2 has
O Tabary et al.
Journal of immunology (Baltimore, Md. : 1950), 164(6), 3377-3384 (2000-03-08)
Increasing evidence suggests that in airways from cystic fibrosis (CF) patients, inflammation may precede bacterial infection and be related to an endogenous dysregulation of proinflammatory cytokines in airway epithelial cells. Several investigators have reported that, in CF airway fluids, elevated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service