M7281
12-Methyltridecanoic acid
98% (capillary GC)
Synonym(s):
(+)-Isomyristic acid, Isomyristic acid
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C14H28O2
CAS Number:
Molecular Weight:
228.37
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
biological source
synthetic (organic)
Quality Level
Assay
98% (capillary GC)
form
powder
functional group
carboxylic acid
lipid type
saturated FAs
shipped in
ambient
storage temp.
2-8°C
SMILES string
CC(C)CCCCCCCCCCC(O)=O
InChI
1S/C14H28O2/c1-13(2)11-9-7-5-3-4-6-8-10-12-14(15)16/h13H,3-12H2,1-2H3,(H,15,16)
InChI key
YYVJAABUJYRQJO-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Anil Sazak et al.
International journal of systematic and evolutionary microbiology, 62(Pt 4), 960-965 (2011-06-15)
A novel actinobacterial strain, A4029T, isolated from arid soil of Abuja, Nigeria, and provisionally assigned to the genus Actinoplanes, was subjected to a polyphasic taxonomic study. 16S rRNA gene sequence similarity studies showed that strain A4029T belonged to the genus
W Shaikh et al.
Pakistan journal of pharmaceutical sciences, 4(1), 55-61 (1991-01-01)
A marine brown alga, Padina tetrastmmatica Hauck, collected from Buleji near Karachi, has been investigated phycochemically. Five saturated fatty acids, i.e. isomyristic, palmitic, margaric, stearic and arachidic, three unsaturated fatty acids, i.e. palmitoleic, oleic and tetradecatrienoic have been detected and
Hyo-Jin Lee et al.
International journal of systematic and evolutionary microbiology, 62(Pt 4), 856-859 (2011-05-31)
Two actinobacterial strains, JR-43T and JR-4, were isolated from bamboo (Sasa borealis) rhizosphere soil. The isolates produced grey aerial mycelium and a yellow soluble pigment on ISP 4. Microscopic observation revealed that strains JR-43T and JR-4 produced rectiflexibiles spore chains
A Ishida-Okawara et al.
The Journal of antibiotics, 44(5), 524-532 (1991-05-01)
We found inhibitors, designated aseanostatins P1 and P5, against myeloperoxidase (MPO) release from human polymorphonuclear leukocytes (PMN). Aseanostatins were extracted from an actinomycete isolated in Thailand and purified by a series of column chromatography of charcoal and silica gel, and
William F Wood
Journal of chemical ecology, 30(2), 479-482 (2004-04-29)
Using GC-MS analysis, 11 major volatile compounds were found in the preorbital gland secretion from a female sika deer, Cervus nippon. These compounds are the C14 through C18 straight-chain fatty acids, (ZZ)-9,12-octadecadienoic acid, 12-methyltridecanoic acid, 13-methyltetradecanoic acid, 14-methylpentadecanoic acid, 14-methylhexadecanoic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service