Skip to Content
Merck
All Photos(1)

Key Documents

M6036

Sigma-Aldrich

Myoglobin from human heart

≥95% (SDS-PAGE), buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61
Pricing and availability is not currently available.

biological source

human heart

Quality Level

Assay

≥95% (SDS-PAGE)

form

buffered aqueous glycerol solution

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... MB(4151)

General description

Myoglobin is a heme-related, low-molecular-weight protein. It is mainly found in cardiac and skeletal muscle.[1]

Application

Myoglobin from human heart has been used as a negative control in monitoring isoAsp formation during storage.[2] It has also been used as an antigen in the study to identify self-antigens recognized by serum autoantibodies from unimmunized mice strains.[3]
Myoglobin from human heart was used to study the interactions of a platinum (II) 9-aminoacridine complex with proteins and oligonucleotides by ESI-TOF MS.[4] It was also used in the evaluation of plasma myoglobin using the Stratus CS fluorometer.[5]

Biochem/physiol Actions

Myoglobin may help to increase diffusion and also stores O2 in oxidative muscle.[6] It can momentarily act as an oxygen reservoir to supply oxygen when there is inadequate blood oxygen delivery at the time of intense muscular activity.[7]

Physical form

Solution in 20 mM Tris-HCl, 1 mM EDTA and 50% glycerol, pH 8.5, 2 mg protein per mL (Lowry)

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iron metabolism and its disorders
Clinical biochemistry of domestic animals, 259-285 (2008)
A High-Throughput Bioluminescent Assay to Monitor the Deamidation of Asparagine and Isomerization of Aspartate Residues in Therapeutic Proteins and Antibodies
Hsiao K, et al.
Journal of Pharmaceutical Sciences, 106(6), 1528-1537 (2017)
Signe Helbo et al.
PloS one, 9(5), e97012-e97012 (2014-06-01)
The discovery that cysteine (Cys) S-nitrosation of trout myoglobin (Mb) increases heme O2 affinity has revealed a novel allosteric effect that may promote hypoxia-induced nitric oxide (NO) delivery in the trout heart and improve myocardial efficiency. To better understand this
Diagnosis of Acute Myocardial Infarction
Cardiac Intensive Care (Second Edition) (2010)
Autoantibody patterns in diabetes-prone NOD mice and in standard C57BL/6 mice
Quintana F J and Cohen I R
Journal of Autoimmunity, 17(3), 191-197 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service