Skip to Content
Merck
All Photos(1)

Documents

M4696

Sigma-Aldrich

Anti-Matrix Metalloproteinase-1 antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-MMP-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 2-15 μg/mL
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP1(4312)

Specificity

The antibody reacts with reduced and non-reduced MMP-1 and recognizes the proform and active forms of MMP-1.

Immunogen

purified, CHO cell-derived, recombinant human MMP-1 (interstitial collagenase).

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Corinne Granger et al.
Dermatology and therapy, 10(6), 1315-1329 (2020-09-12)
Extrinsic factors, such as solar radiation and urban pollution, cause damage that alters the structure, function and appearance of skin. The aim of this study was to determine the ability of a night cream containing melatonin, carnosine and Helichrysum italicum extract
Magnus S Ågren et al.
European journal of cell biology, 94(1), 12-21 (2014-12-03)
Tumor necrosis factor (TNF)-α induces matrix metalloproteinases (MMPs) that may disrupt skin integrity. We have investigated the effects and mechanisms of exogenous TNF-α on collagen degradation by incubating human skin explants in defined serum-free media with or without TNF-α (10ng/ml)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service