Skip to Content
Merck
All Photos(2)

Key Documents

M4251

Sigma-Aldrich

3-Methoxytyramine hydrochloride

≥95.5%, crystalline

Synonym(s):

3-MT, 3-Methoxy-4-hydroxyphenethylamine hydrochloride, 4-(2-Aminoethyl)-2-methoxyphenol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3-4-(OH)CH2CH2NH2·HCl
CAS Number:
Molecular Weight:
203.67
Beilstein:
3631283
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95.5%

form

crystalline

drug control

Home Office Schedule 1

color

white to light brown

mp

213-215 °C (lit.)

storage temp.

−20°C

SMILES string

Cl[H].COc1cc(CCN)ccc1O

InChI

1S/C9H13NO2.ClH/c1-12-9-6-7(4-5-10)2-3-8(9)11;/h2-3,6,11H,4-5,10H2,1H3;1H

InChI key

AWRIOTVUTPLWLF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Major metabolite of dopamine; product of catechol O-methyltransferase.

Features and Benefits

This compound is featured on the Dopamine and Norepinephrine Metabolism page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moon Chul Jung et al.
Analytical chemistry, 78(6), 1761-1768 (2006-03-16)
The photoluminescence-following electron-transfer (PFET) technique, developed in our laboratory, is a sensitive chromatographic detection method for oxidizable analytes. Because the oxidations are homogeneous, the technique avoids the problem of electrode fouling. A liquid-phase oxidant reacts with the electrochemically active analytes
Wilhelmina H A de Jong et al.
The Journal of clinical endocrinology and metabolism, 94(8), 2841-2849 (2009-07-02)
Measurements of the 3-O-methylated metabolites of catecholamines [metanephrines (MNs)] in plasma or urine are recommended for diagnosis of pheochromocytoma. It is unclear whether these tests are susceptible to dietary influences. The aim of the study was to determine the short-term
Marc Morissette et al.
Journal of neuroscience research, 87(7), 1634-1644 (2008-12-31)
A higher prevalence and incidence of Parkinson's disease is observed in men, and beneficial motor effects of estrogens are observed in parkinsonian women. In rodents, an effect of estradiol on dopamine systems is documented, whereas much less is known in
Graeme Eisenhofer et al.
Clinical chemistry, 57(3), 411-420 (2011-01-26)
Pheochromocytomas are rare catecholamine-producing tumors derived in more than 30% of cases from mutations in 9 tumor-susceptibility genes identified to date, including von Hippel-Lindau tumor suppressor (VHL); succinate dehydrogenase complex, subunit B, iron sulfur (Ip) (SDHB); and succinate dehydrogenase complex
Amal Alachkar et al.
Neuroscience research, 67(3), 245-249 (2010-03-23)
The ability of l-3,4-dihydroxyphenylalanine (l-DOPA), l-DOPA-methyl ester and their major metabolites, dopamine, dihydroxyphenylacetic acid (DOPAC), homovanillic (HVA), 3-O-methyldopa and 3-methoxytyramine (3-MT) to bind to alpha(2) adrenergic and D1 and D2 dopamine receptors was assessed by radioligand binding to cloned human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service