Skip to Content
Merck
All Photos(1)

Documents

M1787

Sigma-Aldrich

Manganese(II) chloride solution

BioReagent, for molecular biology, storage temp.:room temp

Synonym(s):

Manganese chlroide, Manganese(II) chloride, MnCl2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnCl2
CAS Number:
Molecular Weight:
125.84
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.52

grade

for molecular biology

Quality Level

sterility

0.2 μm filtered

product line

BioReagent

form

liquid

concentration

1.00 M±0.01 M

cation traces

heavy metals (as Pb): ≤5 ppm

foreign activity

DNase, Exonuclease; NICKase, Endonuclease; and protease, none detected

storage temp.

room temp

SMILES string

Cl[Mn]Cl

InChI

1S/2ClH.Mn/h2*1H;/q;;+2/p-2

InChI key

GLFNIEUTAYBVOC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Manganese (II) chloride (MnCl2) might play a key role in integrin activation. Manganese is indispensable for brain function and is an essential cofactor for several key enzymes like superoxide dismutase and enzymes involved in neurotransmitter synthesis. Manganese can modulate iron and calcium homeostasis. Manganese is also used in the magnetic resonance imaging to increase contrast while examining neuronal connections and brain cytoarchitecture. High levels of manganese is toxic to central nervous system and causes Parkinson′s like symptoms called as manganism. MnCl2 has been used in reverse transcription reaction mix to enhance the template-switching efficiency.
MnCl2 solution is a source of manganese ions and has a variety of biological uses. In biochemistry, it can be used as a cofactor in restriction enzymes, such as BamH and EcoR V. In molecular biology, Mn2+ can be substituted for Mg2+ as a divalent metal ion to facilitate DNase I activity in presence of Ca2+.

Application

Manganese (II) chloride solution has been used:
  • in manganese enhanced magnetic resonance imaging (MEMRI) to study neuronal disease and manganese toxicity
  • in high ionic strength reaction mixture to determine RNA polymerase I and II activity
  • in reverse transcription reaction mix as a source of Mn2+ ions

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - STOT RE 2

Target Organs

Brain

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro Assessment of RNA Polymerase I Activity
Govoni M
Oncogene (2016)
Molecular Cloning: A Laboratory Manual
Sambrook, J. and Russel, D.W.
Molecular Cloning: A Laboratory Manual, 1, A4-A4 (2001)
Highly multiplexed and strand-specific single-cell RNA 5′ end sequencing
Islam S, et al.
Nature Protocols, 7(5), 813-813 (2012)
Manganese enhanced MRI in rat hippocampus: A correlative study with synchrotron X-ray microprobe
Daoust A, et al.
Neuroimage, 64, 10-18 (2013)
Syndecan-1 regulates cell migration and fibronectin fibril assembly
Stepp MA, et al.
Experimental Cell Research, 316(14), 2322-2339 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service