Skip to Content
Merck
All Photos(1)

Key Documents

M1195

Sigma-Aldrich

Metolazone

≥98% (HPLC), solid

Synonym(s):

7-Chloro-1,2,3,4-tetrahydro-2-methyl-3-(2-methylphenyl)-4-oxo-6-quinazolinesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16ClN3O3S
CAS Number:
Molecular Weight:
365.83
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: >5 mg/mL

SMILES string

CC1Nc2cc(Cl)c(cc2C(=O)N1c3ccccc3C)S(N)(=O)=O

InChI

1S/C16H16ClN3O3S/c1-9-5-3-4-6-14(9)20-10(2)19-13-8-12(17)15(24(18,22)23)7-11(13)16(20)21/h3-8,10,19H,1-2H3,(H2,18,22,23)

InChI key

AQCHWTWZEMGIFD-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibitor of thiazide-sensitive Na+-Cl- cotransporter; antihypertensive; moderate "loop" diuretic.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nigel Sparrow et al.
European journal of heart failure, 5(3), 355-361 (2003-06-12)
The National Service Framework (NSF) sets standards for the management of heart failure in the UK. Loop diuretics are commonly first prescribed in primary care. Some patients taking these drugs have heart failure and may benefit from other treatments including
[Therapy edema: secondary effect without effect!].
B Truniger
Schweizerische medizinische Wochenschrift, 129(46), 1808-1808 (1999-12-22)
S C Hebert et al.
Kidney international, 49(6), 1638-1641 (1996-06-01)
Recently the molecular identification of the major electroneutral sodium-potassium-chloride entry mechanisms present on apical membranes of distal nephron segments of the mammalian kidney, on basolateral membranes of many non-renal epithelial cells and on certain non-epithelial tissues has been achieved. These
Jens Rosenberg et al.
Cardiovascular drugs and therapy, 19(4), 301-306 (2005-09-29)
Metolazone is a potent thiazide-like diuretic. It is recommended in severe congestive heart failure (HF). We conducted a review of the existing literature and found that the available information on the use of metolazone in HF is based on studies
A Monroy et al.
American journal of physiology. Renal physiology, 279(1), F161-F169 (2000-07-15)
The thiazide-sensitive Na(+)-Cl(-) cotransporter (TSC) is the major pathway for salt reabsorption in the apical membrane of the mammalian distal convoluted tubule. When expressed in Xenopus laevis oocytes, rat TSC exhibits high affinity for both cotransported ions, with the Michaelis-Menten

Articles

Voltage-gated sodium channels are present in most excitable cell membranes and play an important role in generating action potentials.

Voltage-gated sodium channels are present in most excitable cell membranes and play an important role in generating action potentials.

Voltage-gated sodium channels are present in most excitable cell membranes and play an important role in generating action potentials.

Voltage-gated sodium channels are present in most excitable cell membranes and play an important role in generating action potentials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service