Skip to Content
Merck
All Photos(2)

Documents

M1177

Sigma-Aldrich

Myriocin

from Mycelia sterilia, ≥98% (HPLC), powder, immunosuppressant

Synonym(s):

(2S,3R,4R,6E)-2-Amino-3,4-dihydroxy-2-(hydroxymethyl)-14-oxo-6-eicosenoic acid, ISP-I, Thermozymocidin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H39NO6
CAS Number:
Molecular Weight:
401.54
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

product name

Myriocin from Mycelia sterilia, ≥98% (HPLC), powder

Quality Level

Assay

≥98% (HPLC)

form

powder

color

off-white

solubility

methanol: 2 mg/mL

storage temp.

2-8°C

SMILES string

CCCCCCC(=O)CCCCCC\C=C\C[C@@H](O)[C@H](O)[C@@](N)(CO)C(O)=O

InChI

1S/C21H39NO6/c1-2-3-4-10-13-17(24)14-11-8-6-5-7-9-12-15-18(25)19(26)21(22,16-23)20(27)28/h9,12,18-19,23,25-26H,2-8,10-11,13-16,22H2,1H3,(H,27,28)/b12-9+/t18-,19+,21+/m1/s1

InChI key

ZZIKIHCNFWXKDY-GNTQXERDSA-N

Application

Myriocin from Mycelia sterilia has been used as an inhibitor of sphingolipid or ceramide biosynthesis in various studies.

Biochem/physiol Actions

Myriocin from Mycelia sterilia is a fungal metabolite with potent immunosuppressant activity. It inhibits serine palmitoyltransferase at picomolar concentrations blocking synthesis of ceramide, a precursor of sphingomyelin and glycosphingolipids. It disrupts substratum adhesion of melanoma cells. It has been suggested that its immunosuppressant activity in the cytotoxic T-cell line CTTL-2 is due to apoptosis induction.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quan-Jiang Zhang et al.
Diabetes, 61(7), 1848-1859 (2012-05-16)
Vascular dysfunction that accompanies obesity and insulin resistance may be mediated by lipid metabolites. We sought to determine if vascular ceramide leads to arterial dysfunction and to elucidate the underlying mechanisms. Pharmacological inhibition of de novo ceramide synthesis, using the
John R Ussher et al.
PloS one, 7(5), e37703-e37703 (2012-05-26)
Diet-induced obesity (DIO) leads to an accumulation of intra-myocardial lipid metabolites implicated in causing cardiac insulin resistance and contractile dysfunction. One such metabolite is ceramide, and our aim was to determine the effects of inhibiting de novo ceramide synthesis on
Fernando Martínez-Montañés et al.
Journal of lipid research, 57(11), 2040-2050 (2016-11-03)
Long-chain bases (LCBs) are the precursors to ceramide and sphingolipids in eukaryotic cells. They are formed by the action of serine palmitoyl-CoA transferase (SPT), a complex of integral membrane proteins located in the endoplasmic reticulum. SPT activity is negatively regulated
Cigarette smoke increases cardiomyocyte ceramide accumulation and inhibits mitochondrial respiration
Tippetts TS, et al.
BMC Cardiovascular Disorders, 14(1), 165-165 (2014)
Maija Ruuth et al.
European heart journal, 39(27), 2562-2573 (2018-07-10)
Low-density lipoprotein (LDL) particles cause atherosclerotic cardiovascular disease (ASCVD) through their retention, modification, and accumulation within the arterial intima. High plasma concentrations of LDL drive this disease, but LDL quality may also contribute. Here, we focused on the intrinsic propensity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service