All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H21N3O5S
CAS Number:
Molecular Weight:
307.37
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Product Name
Met-Ala-Ser,
Assay
≥97% (TLC)
Quality Level
form
powder
color
white
storage temp.
−20°C
SMILES string
CSCC[C@H](N)C(=O)N[C@@H](C)C(=O)N[C@@H](CO)C(O)=O
InChI
1S/C11H21N3O5S/c1-6(9(16)14-8(5-15)11(18)19)13-10(17)7(12)3-4-20-2/h6-8,15H,3-5,12H2,1-2H3,(H,13,17)(H,14,16)(H,18,19)/t6-,7-,8-/m0/s1
InChI key
WXHHTBVYQOSYSL-FXQIFTODSA-N
Amino Acid Sequence
Met-Ala-Ser
Application
Met-Ala-Ser has been used as a tripeptide in the OH radical-induced oxidation reaction and analysis by high performance liquid chromatography-diode array detector-fluorescence detector (HPLC-DAD-FLD).
Biochem/physiol Actions
Met-Ala-Ser (MAS) is used to complex with peptide deformylase(s) (PDF) to help delineate mechanisms of recognition of mimic-peptides by N-terminal cleaved expression peptide.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A Becker et al.
Nature structural biology, 5(12), 1053-1058 (1998-12-10)
Eubacterial proteins are synthesized with a formyl group at the N-terminus which is hydrolytically removed from the nascent chain by the mononuclear iron enzyme peptide deformylase. Catalytic efficiency strongly depends on the identity of the bound metal. We have determined
Ki Hyun Nam et al.
Biochemical and biophysical research communications, 381(4), 630-633 (2009-03-03)
PDF (peptide deformylase) plays a critical role in the production of mature proteins by removing the N-formyl polypeptide of nascent proteins in the prokaryote cell system. This protein is essential for bacterial growth, making it an attractive target for the
Ren Miao et al.
Rapid communications in mass spectrometry : RCM, 19(8), 1031-1040 (2005-03-16)
Reactions of two platinum(II) complexes, cis-[Pt(NH3)2(H2O)2]2+ (Pt1) and cis-[Pt(en)(H2O)2]2+ (Pt2), with several sulfur-containing peptides, have been investigated by electrospray ionization mass spectrometry (ESI-MS) and tandem mass spectrometry (MS/MS). The species produced in the reactions were detected with ESI-MS, and MS/MS
Release of free amino acids upon oxidation of peptides and proteins by hydroxyl radicals
Liu F, et al.
Analytical and Bioanalytical Chemistry, 409(9), 2411-2420 (2017)
Qing Tang et al.
The Journal of cell biology, 214(2), 167-179 (2016-07-20)
Myo51, a class V myosin in fission yeast, localizes to and assists in the assembly of the contractile ring, a conserved eukaryotic actomyosin structure that facilitates cytokinesis. Rng8 and Rng9 are binding partners that dictate the cellular localization and function
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service