Skip to Content
Merck
All Photos(1)

Key Documents

M0269

Sigma-Aldrich

Methotrexate−Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
13111023
NACRES:
NA.56

biological source

plant

Quality Level

form

saline suspension

extent of labeling

2-7 mg per mL

technique(s)

affinity chromatography: suitable

matrix

cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

carboxyl

matrix spacer

8 atoms

suitability

suitable for chromatography

storage temp.

2-8°C

Application

Methotrexate-agarose is used in protein chromatography, affinity chromatography, metabolic pathways and specialty resins. Methotrexate-agarose has been used to investigate the toxicity mechanism of mortality in adult buffalo flies caused by ingestion of folate analogues. Methotrexate-agarose has also been used to study the purification, cloning, and functional expression of dihydroneopterin triphosphate 2′-epimerase from Escherichia coli.

Physical form

Suspension in 1.0 M NaCl containing preservative

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazumi Urakawa et al.
European journal of pharmacology, 435(2-3), 237-244 (2002-02-01)
N-[[4-[(2,4-diaminopteridin-6-yl)methyl]-3,4-dihydro-2H-1,4-benzothiazin-7-yl]-carbonyl]-L-homoglutamic acid (MX-68), a derivative of methotrexate, was chemically designed to resist polyglutamation and to have a high affinity for dihydrofolate reductase, in an attempt to reduce the side effects of methotrexate. We confirmed that MX-68 did not undergo polyglutamation
T Endo et al.
The EMBO journal, 7(4), 1153-1158 (1988-04-01)
We have purified milligram amounts of an importable mitochondrial precursor protein [the presequence of yeast cytochrome oxidase subunit IV fused to mouse dihydrofolate reductase (DHFR)]. This has made it possible, for the first time, to perform detailed studies on the
C Ahn et al.
The Journal of biological chemistry, 272(24), 15323-15328 (1997-06-13)
Dihydroneopterin triphosphate (H2NTP) 2'-epimerase from Escherichia coli catalyzes the epimerization of H2NTP to dihydromonapterin triphosphate (H2MTP). The enzyme was purified 954-fold to apparent homogeneity by a combination of ammonium sulfate fractionation and column chromatography of Cibacron blue 3GA dye ligand
H J Chung et al.
Biochimica et biophysica acta, 1524(2-3), 183-188 (2000-12-13)
Tetrahydrobiopterin (BH4)-glucoside was identified from Synechococcus sp. PCC 7942 by HPLC analysis and the enzymatic activity of a glycosyltransferase producing the compound from UDP-glucose and BH4. The novel enzyme, named UDP-glucose:BH4 glucosyltransferase, has been purified 846-fold from the cytosolic fraction
M Iwakura et al.
Protein engineering, 5(8), 791-796 (1992-12-01)
A single polypeptide chain containing two dihydrofolate reductase (DHFR) sequences from Escherichia coli was constructed to determine if a repeat sequence fusion protein could be expressed in an active form. The possibility that intersequence interactions could play a significant role

Articles

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

Biofiles reviews innovative technologies for cancer research, reflecting the complexity of the disease.

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service