Skip to Content
Merck
All Photos(1)

Key Documents

L9647

Sigma-Aldrich

Limonin

from citrus seeds, >90% (HPLC)

Synonym(s):

Citrolimonin, Dictamnolactone, Evodin, Limonoic acid 3,19:16,17 dilactone, Obaculactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H30O8
CAS Number:
Molecular Weight:
470.51
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

biological source

citrus seeds

Quality Level

Assay

>90% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

CC1(C)O[C@H]2CC(=O)OC[C@]23[C@H]4CC[C@@]5(C)[C@@H](OC(=O)[C@H]6O[C@@]56[C@]4(C)C(=O)C[C@@H]13)c7ccoc7

InChI

1S/C26H30O8/c1-22(2)15-9-16(27)24(4)14(25(15)12-31-18(28)10-17(25)33-22)5-7-23(3)19(13-6-8-30-11-13)32-21(29)20-26(23,24)34-20/h6,8,11,14-15,17,19-20H,5,7,9-10,12H2,1-4H3/t14-,15-,17-,19-,20+,23-,24-,25+,26+/m0/s1

InChI key

KBDSLGBFQAGHBE-MSGMIQHVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Limonin also known as citrolimonin and evodin, is a secondary plant metabolite present in citrus seeds at high concentrations. It is a derivative of limonoids.
Limonin may be used in assays to quantify limonin in citrus products. It may also be used in other assays as a control, standard, or competitor or in studies on the physiology, metabolism, and biosynthesis of limonin.

Application

Limonin has been used:
  • to treat human peripheral blood mononuclear cells (PBMCs) to test its effect on human immunodeficiency virus (HIV) replication
  • to test its effects as an anti-osteoporosis agent in osteoblastogenesis activity in MC3T3-E1 cells and ovariectomized (OVX) rat model
  • as a reference standard in high-performance liquid chromatography (HPLC) system

Biochem/physiol Actions

Limonin exerts its anti-inflammatory activity by downregulating signal transducer and activator of transcription 3/ microRNA-214 (STAT3/miR-214) signaling pathway in Ulcerative colitis (UC) condition. It is an anti-carcinogenic agent, involved in the down-regulation Wingless-related integration site (Wnt) signaling pathway component in in vitro studies.
Limonin is a triterpenoid aglycone that is a bitter principle of citrus fruits. It has anti-proliferative, proapoptotic activity on several cancer cell lines and inhibits azoxymethane-induced colon cancer in rats. It also inhibits HIV-1 replication in culturedf monocytes, macrophages, and mononuclear cells, perhaps by inhibition of HIV-1 protease activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahmoud Zaki El-Readi et al.
European journal of pharmacology, 626(2-3), 139-145 (2009-09-29)
P-glycoprotein (P-gp), a membrane transporter encoded by the MDR1 gene in human cells, mediates drug efflux from cells and plays a major role in causing multidrug resistance; which is one of the most accepted mechanisms for failure of chemotherapy in
Shunming Fan et al.
Molecules (Basel, Switzerland), 24(20) (2019-10-17)
Limonin is a natural tetracyclic triterpenoid compound, which widely exists in Euodiarutaecarpa (Juss.) Benth., Phellodendronchinense Schneid., and Coptischinensis Franch. Its extensive pharmacological effects have attracted considerable attention in recent years. However, there is no systematic review focusing on the pharmacology
Kotamballi N Chidambara Murthy et al.
Journal of agricultural and food chemistry, 59(6), 2314-2323 (2011-02-23)
The current study was an attempt to elucidate the mechanism of human colon cancer cell proliferation inhibition by limonin and limonin glucoside (LG) isolated from seeds of Citrus reticulata. The structures of purified compounds were confirmed by NMR and quantified
K Langeswaran et al.
Journal of natural science, biology, and medicine, 4(1), 126-133 (2013-05-02)
The role of limonin as potent anti carcinogenic, apoptosis and chemotherapeutic agents has been supported by limited studies. In this study, limonin is identified as a potent anti proliferative agent against human hepatoma HepG2 cells based on the cell viability
Yang Zhao et al.
Journal of pharmaceutical and biomedical analysis, 48(4), 1230-1236 (2008-10-22)
A simple, sensitive and accurate HPLC-DAD method was developed for simultaneous determination of wuchuyuamide-I, quercetin, limonin, evodiamine and rutaecarpine in Evodia rutaecarpa that has been widely used as one of the traditional Chinese medicines (TCMs). Chromatographic separations were performed on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service