Skip to Content
Merck
All Photos(1)

Documents

L9250

Sigma-Aldrich

Leu-Ala hydrate

Bulk package

Synonym(s):

L-Leucyl-L-alanine hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(NH2)CONHCH(CH3)COOH · xH2O
CAS Number:
Molecular Weight:
202.25 (anhydrous basis)
Beilstein:
1726165
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Quality Level

Assay

≥98% (TLC)

form

powder

technique(s)

ligand binding assay: suitable

color

white

storage temp.

2-8°C

SMILES string

O.CC(C)C[C@H](N)C(=O)N[C@@H](C)C(O)=O

InChI

1S/C9H18N2O3.H2O/c1-5(2)4-7(10)8(12)11-6(3)9(13)14;/h5-7H,4,10H2,1-3H3,(H,11,12)(H,13,14);1H2/t6-,7-;/m0./s1

InChI key

FTDFAGANYJVHDA-LEUCUCNGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Leucylalanine (Leu-Ala) is useful to produce non-electrolytic triorganotin (IV) derivatives (R3Sn(HL)) to study the models of metal-protein interactions.

Application

Leu-Ala hydrate has been used as a substrate in Tris-hydrochloride (HCl) buffer to analyze the leucine-alanine peptidase (LAP) activity in larvae.
Leucylalanine (Leu-Ala) is use to make non-electrolytic triorganotin(IV) derivatives (general formulae R3Sn(HL)) to study models of metal-protein interactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New triorganotin (IV) derivatives of dipeptides as models for metal-protein interactions: synthesis, structural characterization and biological studies.
Nath M, Pokharia S, Eng G, Song X, Kumar A.
Spectrochimica Acta Part A: Molecular Spectroscopy, 63, 66-75 (2006)
David S Milner et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(12), 5613-5622 (2019-03-08)
Many microbes acquire metabolites in a "feeding" process where complex polymers are broken down in the environment to their subunits. The subsequent uptake of soluble metabolites by a cell, sometimes called osmotrophy, is facilitated by transporter proteins. As such, the
Marie Terpager et al.
Journal of receptor and signal transduction research, 29(5), 235-245 (2009-09-15)
7TM receptors are easily fused to proteins such as G proteins and arrestin but because of the fact that their terminals are found on each side of the membrane they cannot be joined directly in covalent dimers. Here, we use
L P Biały et al.
Folia histochemica et cytobiologica, 40(2), 135-136 (2002-06-12)
We have used the dipeptide Leu-Ala in an attempt to prevent the formation of ubiquitin-protein conjugates in U937 cells by inhibition of cellular E3 enzymes (ubiquitin ligases). Proteasome inhibitors induce the formation of perinuclear aggregates of ubiquitinated proteins and proteasomes
Arun K Ghosh et al.
Journal of medicinal chemistry, 50(10), 2399-2407 (2007-04-17)
Structure-based design and synthesis of a number of potent and selective memapsin 2 inhibitors are described. These inhibitors were designed based upon the X-ray structure of memapsin 2-bound inhibitor 3 that incorporates methylsulfonyl alanine as the P2-ligand and a substituted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service