Skip to Content
Merck
All Photos(2)

Key Documents

L8146

Sigma-Aldrich

Lectin from Ulex europaeus (gorse, furze)

Lectin from Ulex europaeus (gorse, furze)
1 of 1 reviewers received a sample product or took part in a promotion

peroxidase conjugate, lyophilized powder

Synonym(s):

Ulex europaeus agglutinin, UEA-I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

Ulex europaeus

Quality Level

conjugate

peroxidase conjugate

Assay

70.00-90.00% (Lowry)

form

lyophilized powder

potency

<40 μg/mL agglutination activity

peroxidase activity

≥15 units/mg protein

composition

Protein, ~80% Lowry

technique(s)

western blot: suitable

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

General Western Blot Protocol:
  • Glycoprotein sample size: 500ng
  • Lectin Concentration: 0.1ug/ml

  1. Load samples at 500 ng of glycoprotein per lane
  2. Run 4-20% Bis-Tris SDS page gel
  3. Transfer gel to a PVDF membrane
  4. Block membrane for 1 hr at RT with RIPA buffer (R0278 Sigma)
  5. Incubate HRP lectin at 0.1ug/ml with RIPA buffer for 2 hours at RT
  6. Wash membrane 5 x 5 minutes with 25ml RIPA buffer
  7. Detect using chemiluminescent substrate (CPS1-120)

Biochem/physiol Actions

Ulex europaeus agglutinin has anti-H blood group specificity. There are two types of lectin: UEA I has an affinity for L-fucose and UEA II has an affinity for N,N′-diacetylchitobiose. The mol. wt. of UEA I was initially found to be 170,000 daltons. Later reports indicate that UEA I may form aggregates at neutral and basic pH and that the correct mol. wt. is 68,000 daltons.

Packaging

Package size based on protein content.

Linkage

Conjugates are prepared from affinity purified lectin.

Unit Definition

One unit will form 1.0 mg purpurogallin in 20 sec from pyrogallol at pH 6.0 at 20 °C.

Physical form

Contains sodium citrate buffer salts

Preparation Note

Prepared from peroxidase, Type VI (P8375), using the two-step glutaraldehyde method of Avrameas and Ternyck.[1]
Repurified by affinity chromatography after conjugation.

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1 mg/ml solution. This activity is the lowest concentration to agglutinate a 2% suspension of human blood group O erythrocytes after 1 hour incubation at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peroxidase labelled antibody and Fab conjugates with enhanced intracellular penetration.
S Avrameas et al.
Immunochemistry, 8(12), 1175-1179 (1971-12-01)
Nadine Mikuda et al.
The Journal of pathology, 251(2), 160-174 (2020-03-30)
The IκB kinase (IKK)-NF-κB signaling pathway plays a multifaceted role in inflammatory bowel disease (IBD): on the one hand, it protects from apoptosis; on the other, it activates transcription of numerous inflammatory cytokines and chemokines. Although several murine models of
K Nozaki et al.
Journal of neurology, neurosurgery, and psychiatry, 80(8), 904-908 (2009-02-26)
To study the clinical and pathological correlations of neuromuscular patients with a high aldolase and normal creatine kinase (CK) in serum at presentation or during a symptomatic exacerbation. Records and muscle biopsies were retrospectively reviewed in a consecutive series of
Xiaohu Fan et al.
Human gene therapy, 21(7), 877-890 (2010-02-19)
The ABO histo-blood group system is the most important antigen system in transplantation medicine, yet no small animal model of the ABO system exists. To determine the feasibility of developing a murine model, we previously subcloned the human alpha-1,2-fucosyltransferase (H-transferase
F Bucardo et al.
Zoonoses and public health, 63(8), 600-607 (2016-05-14)
Information about porcine norovirus (PoNoV), genetically similar to human NoV (HuNoV), is limited from rural areas where household-raised pigs are heavily exposed to faecal material which could facilitate transmission. Histo-blood group antigens (HBGAs) are known susceptibility factors to NoV in

Articles

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. Dublin, Ireland
    • Review 1
    • Votes 0
    5 out of 5 stars.

    It worked perfectly.

    I have been struggled with high background using other companies Ulex, but this one just worked perfectly since first time.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service