Skip to Content
Merck
All Photos(1)

Key Documents

L3888

Sigma-Aldrich

D-Lactic Dehydrogenase from Lactobacillus leichmannii

lyophilized powder, 150-500 units/mg protein

Synonym(s):

(R)-Lactate:NAD+ oxidoreductase, D-LDH

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bacterial (Lactobacillus leichmannii)

Quality Level

form

lyophilized powder

specific activity

150-500 units/mg protein

composition

Protein, ~50% biuret

foreign activity

Malic dehydrogenase <0.5% of base activity

storage temp.

−20°C

Related Categories

General description

Research area: Cell Signaling
Lactate Dehydrogenase (LDH) is classified as an oxidoreductase and is found in various organisms, including both plants and animals. LDH is widely distributed across all tissues, with high concentrations in muscle, kidney, and liver. The genes encoding LDH are LDHA, LDHB, LDHC, and LDHD. The D-isomer is produced by LDHD. There are two types of D-LDHs: NAD-dependent D-LDHs and FAD-dependent D-LDHs.

Application

D-Lactic Dehydrogenase from Lactobacillus leichmannii has been used:
  • in lactate dehydrogenase activity for testing the chaperone activity of proteins
  • to test the kinase activities of purified thiamine monophosphate(ThiM)
  • in NADH-coupled steady-state ATPase assay
  • to determine cellular lactate
In the food industry, the primary catalysis is coupled to conversion of NADH and H+ to NAD+ with diaphorase coupled with converting the non-fluorescent resazurin to the highly fluorescent substance resorufin to measure the content of D-lactate in food products.

Biochem/physiol Actions

D-lactic dehydrogenase catalyzes the conversion of pyruvate into D-lactate, with oxidation of NADH to NAD+. D-lactic dehydrogenase can also catalyze the reverse reaction, conversion of D-lactate into pyruvate with reduction of NAD+ to NADH.
It acts as a crucial checkpoint in gluconeogenesis and DNA metabolism. Elevated levels of LDH in the blood have been observed in various conditions, including heart attacks, cancers, liver disease, muscle trauma, anemia, bone fractures, and infections such as encephalitis, human immunodeficiency virus(HIV), and meningitis. LDH also serves as a non-specific marker of tissue turnover, which is a normal metabolic process. Additionally, reduced D-LDH activity has been found in case of mutations in LDHD found in patients with D-lactic acidosis.

Unit Definition

D-lactic dehydrogenase catalyzes the conversion of pyruvate into D-lactate, with oxidation of NADH to NAD+. D-lactic dehydrogenase can also catalyze the reverse reaction, conversion of D-lactate into pyruvate with reduction of NAD+ to NADH.
One unit will reduce 1.0 μmole of pyruvate to D-lactate per min at pH 7.0 at 25 °C.

Physical form

Lyophilized powder containing phosphate buffer salts

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lactate dehydrogenase D is a general dehydrogenase for D-2-hydroxyacids and is associated with D-lactic acidosis
Shan J, et al.
Nature Communications, 14, 6638-6638 (2023)
Biochemistry, Lactate Dehydrogenase
Farhana A and Lappin SL
StatPearls [Internet] (2023)
Structural Insights into Mdn1, an Essential AAA Protein Required for Ribosome Biogenesis
Chen Z, et al.
Cell, 175(3), 822?834-822?834 (2018)
Cysteine Deprivation Targets Ovarian Clear Cell Carcinoma Via Oxidative Stress and Iron?Sulfur Cluster Biogenesis Deficit
Novera W, et al.
Antioxidants & Redox Signaling, 33(17), 1191?1208-1191?1208 (2020)
A Novel Method for Assessing the Chaperone Activity of Proteins
Hristozova N, et al.
PLoS ONE, 11(8), e0161970-e0161970 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service