Skip to Content
Merck
All Photos(2)

Key Documents

L3001

Sigma-Aldrich

Lipase from wheat germ

Type I, lyophilized powder, 5-15 units/mg solid

Synonym(s):

Triacylglycerol acylhydrolase, Triacylglycerol lipase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

wheat (germ)

type

Type I

form

lyophilized powder

specific activity

5-15 units/mg solid

UniProt accession no.

storage temp.

2-8°C

InChI

1S/C11H9N3O2.Na/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11;/h1-7,16H,(H,12,14);/q;+1/b13-9-;

InChI key

QWZUIMCIEOCSJF-CHHCPSLASA-N

Gene Information

wheat ... lipase(543006)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Lipases belong to the class of hydrolases. Microbes of plant and animal basis mainly produce lipase enzyme. It can also be produced by marine organisms.

Application

Lipase from wheat germ has been used:
  • to prepare enzyme solution for the biodegradability test
  • to treat diacetates of Mep-Xylp, to generate monoacetylated xylosides
  • to test its efficacy in selenium speciation analysis and to determine the contribution that impurities could make to Se determination in real samples

Biochem/physiol Actions

Lipases are used in detergent, food, leather, pharmaceutical and dairy industries. It has several characteristics like substrate specificity, stereospecificity and regioselectivity. It also participates in various bioconversion reactions like, hydrolysis, esterification, acidolysis and aminolysis.
Tri-, di-, and monoglycerides are hydrolyzed (in decreasing order of rate).

Quality

Contains acid phosphatase.

Unit Definition

One unit will hydrolyze 1.0 microequivalent of fatty acid from a triglyceride in 1 hr at pH 7.4 at 37 °C. (This is equivalent to approx. 10 microliters of CO2 in 30 minutes.)

Preparation Note

Prepared essentially by method of Singer, T.P., J. Biol. Chem., 174, 11 (1948).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ondřej Dlouhý et al.
Cells, 10(9) (2021-09-29)
Build-up of the energized state of thylakoid membranes and the synthesis of ATP are warranted by organizing their bulk lipids into a bilayer. However, the major lipid species of these membranes, monogalactosyldiacylglycerol, is a non-bilayer lipid. It has also been
Lipase: An Industrial Enzyme Through Metagenomics
Advances in Food and Nutrition Research, 78, 71-94 (2018)
Ondřej Dlouhý et al.
Cells, 10(9) (2021-09-29)
In Part I, by using 31P-NMR spectroscopy, we have shown that isolated granum and stroma thylakoid membranes (TMs), in addition to the bilayer, display two isotropic phases and an inverted hexagonal (HII) phase; saturation transfer experiments and selective effects of
Preparation and characterization of poly (ethylene glycol)-crosslinked reacetylated chitosans
Dal P A, et al.
Carbohydrate Polymers, 42(2), 201-206 (2000)
Marine fungal and bacterial isolates for lipase production: A comparative study
Advances in Food and Nutrition Research, 78, 71-94 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service