Skip to Content
Merck
All Photos(1)

Key Documents

L2290

Sigma-Aldrich

Anti-LIM Kinase 1 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-LIMK1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

technique(s)

microarray: suitable
western blot: 1:2,000 using a cytosolic fraction of rat brain.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LIMK1(3984)
mouse ... Limk1(16885)
rat ... Limk1(65172)

General description

LIM Domain Kinase 1 (LIMK1) belongs to the LIMK family of serine/threonine protein kinases. LIMK1 shares about 50% sequence identity with LIMK2. LIMK1 contains two N-terminal LIM domains, an internal PDZ (post synaptic density protein (PSD95), Drosophila disc large tumor suppressor (Dlg1), and zonula occludens-1 protein (zo-1)) domain and a C-terminal kinase domain.

Immunogen

Synthetic peptide corresponding to the C-terminus of mouse LIMK1 (amino acids 627-647 or 613-633 of the alternatively spliced forms of LIMK1), conjugated to KLH
This sequence is identical in rat LIMK1 and highly conserved (single amino acid substitution) in human LIMK1. It is not found in the LIMK2 isoform.

Application

Anti-LIM Kinase 1 antibody is suitable for use in microarrays, protein densitometric assays (1:1000 using BPAE cell extracts ) and western blot (1:100, 000 using MDA-MB-231 cell lysates ).

Biochem/physiol Actions

LIMK1 phosphorylates cofilin specifically at Serine 3. This inactivates its F-actin depolymerizing activity and leads to actin reorganization and accumulation of actin filaments. LIMK1 mRNA is highly expressed in the developing nervous system, heart and gut, and in adult brain and spinal cord, suggesting a role for LIMK-1 during neuronal cell differentiation. LIMK1 is localized in the presynaptic nerve ending during the synapse maturation event. LIMK1 also exhibits mitosis-specific activation.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tissue specific characterisation of Lim-kinase 1 expression during mouse embryogenesis
Lindstrom NO, et al.
Gene Expression Patterns, 11(3-4), 221-232 (2011)
Mitosis-specific activation of LIM motif-containing protein kinase and roles of cofilin phosphorylation and dephosphorylation in mitosis
Amano T, et al.
The Journal of biological chemistry, 277(24), 22093-22102 (2002)
Zoe M Goeckeler et al.
The Journal of biological chemistry, 280(38), 33083-33095 (2005-08-02)
This study determined the effects of increased intracellular cAMP and cAMP-dependent protein kinase activation on endothelial cell basal and thrombin-induced isometric tension development. Elevation of cAMP and maximal cAMP-dependent protein kinase activation induced by 10 microm forskolin, 40 microm 3-isobutyl-1-methylxanthine
Patricia Castañeda et al.
Journal of neuroscience research, 93(10), 1476-1491 (2015-05-27)
Chronic stress promotes cognitive impairment and dendritic spine loss in hippocampal neurons. In this animal model of depression, spine loss probably involves a weakening of the interaction between pre- and postsynaptic cell adhesion molecules, such as N-cadherin, followed by disruption
K Ohashi et al.
The Journal of biological chemistry, 275(5), 3577-3582 (2000-02-01)
LIM-kinase 1 (LIMK1) phosphorylates cofilin, an actin-depolymerizing factor, and regulates actin cytoskeletal reorganization. LIMK1 is activated by the small GTPase Rho and its downstream protein kinase ROCK. We now report the site of phosphorylation of LIMK1 by ROCK. In vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service