Skip to Content
Merck
All Photos(4)

Key Documents

L2157

Sigma-Aldrich

Laccase from Rhus vernicifera

greener alternative

crude acetone powder, ≥50 units/mg solid

Synonym(s):

Benzenediol:oxygen oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

form

crude acetone powder

Quality Level

specific activity

≥50 units/mg solid

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

−20°C

SMILES string

[n]1(c(c(cc1C)C=O)C)CC

InChI

1S/C9H13NO/c1-4-10-7(2)5-9(6-11)8(10)3/h5-6H,4H2,1-3H3

InChI key

NWDZDFOKSUDVJV-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in biofuel cell and cellulosic ethanol research. For more information see the article in biofiles and Enzymes for Alternative Energy Research.

Unit Definition

One unit will produce a ΔA530 of 0.001 per min at pH 6.5 at 30 °C in a 3 mL reaction volume using syringaldazine as substrate.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jervian Johnson et al.
3 Biotech, 11(7), 360-360 (2021-07-24)
In this study, hydrolytic and oxidative activities of enzymes isolated from halophilic microbes were characterized and applied for biomass utilization. First, lipase from Micrococcus luteus, and peroxidase and laccase from Pseudoalteromonas phenolica and Pseudoalteromonas peptidolytica were selected and their catalytic
Shanfa Lu et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(26), 10848-10853 (2013-06-12)
Laccases, as early as 1959, were proposed to catalyze the oxidative polymerization of monolignols. Genetic evidence in support of this hypothesis has been elusive due to functional redundancy of laccase genes. An Arabidopsis double mutant demonstrated the involvement of laccases
Ngoc Han Tran et al.
Bioresource technology, 146, 721-731 (2013-08-21)
Many efforts have been made to understand the biodegradation of emerging trace organic contaminants (EOCs) in the natural and engineered systems. This review summarizes the current knowledge on the biodegradation of EOCs while having in-depth discussion on metabolism and cometabolism
Mohammed Sherif et al.
Microbial biotechnology, 6(5), 588-597 (2013-07-03)
Multicopper oxidases can act on a broad spectrum of phenolic and non-phenolic compounds. These enzymes include laccases, which are widely distributed in plants and fungi, and were more recently identified in bacteria. Here, we present the results of biochemical and
Yu-Chan Zhang et al.
Nature biotechnology, 31(9), 848-852 (2013-07-23)
Increasing grain yields is a major focus of crop breeders around the world. Here we report that overexpression of the rice microRNA (miRNA) OsmiR397, which is naturally highly expressed in young panicles and grains, enlarges grain size and promotes panicle

Protocols

Laccase, a biocatalyst, oxidizes aromatic compounds with molecular oxygen reduction, making it useful in various applications.

Laccase, a biocatalyst, oxidizes aromatic compounds with molecular oxygen reduction, making it useful in various applications.

Laccase, a biocatalyst, oxidizes aromatic compounds with molecular oxygen reduction, making it useful in various applications.

Laccase, a biocatalyst, oxidizes aromatic compounds with molecular oxygen reduction, making it useful in various applications.

Questions

1–6 of 6 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What is the pH optimum of Product L2157, Laccase from Rhus vernicifera?

    1 answer
    1. Our own assay is performed at pH 6.5.  One publication (Casella et al., J. Inorg. Biochem., 100, 2127-2139 (2006)) gives pH 6.5 as another value for the laccase from this species, with a range for enzyme activity of 4.3 to 9.

      Helpful?

  3. My solution of Product L2157, Laccase from Rhus vernicifera, isn't clear. Is this normal?

    1 answer
    1. Yes. L2157 is a crude acetone powder, and can contain other proteins, enzymes, and cellular debris.  A suspension may be prepared in 100 mM potassium phosphate buffer, pH 6.5, by homogenization with a hand-held homogenizer for 10 minutes. This preparation may then be centrifuged; the laccase activity will be present in the supernatant.

      Helpful?

  4. How stable are solutions of Product L2157, Laccase from Rhus vernicifera?

    1 answer
    1. Solutions should be prepared fresh, directly before use, as they are not stable as stock solutions.

      Helpful?

  5. What is the molecular weight (MW) of Product L2157, Laccase from Rhus vernicifera?

    1 answer
    1. Spira-Solomon and Solomon, J. Am. Chem. Soc., 109, 6421-6432 (1987) reports estimated molecular weights of 52 kDa (laccase 1), 55 kDa (laccase 2), and 70 kDa (laccase 3), as determined by gel filtration.

      Helpful?

  6. What is the temperature optimum of Product L2157, Laccase from Rhus vernicifera?

    1 answer
    1. Our own assay is performed at 30°C.  One publication (Casella et al., J. Inorg. Biochem., 100, 2127-2139 (2006)) gives 25°C as another value for the same enzyme from the same species.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service