Skip to Content
Merck
All Photos(1)

Key Documents

K4377

Sigma-Aldrich

Monoclonal Anti-Human Kappa Light Chain (Bound and Free) antibody produced in mouse

clone KP-53, ascites fluid

Synonym(s):

Monoclonal Anti-Human Kappa Light Chain (Bound and Free)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

secondary antibodies

clone

KP-53, monoclonal

contains

15 mM sodium azide

technique(s)

indirect ELISA: 1:10,000

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal Anti-Human Kappa Light Chain (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Immunoglobulins G are made up of two identical light chains and two identical heavy chains, that are covalently connected by interchain disulphide bonds. Genes of kappa and lambda constant region are mapped on to human chromosome 2.

Specificity

The antibody is specific for human kappa (κ) light chains. This antibody is reactive with intact IgG (κ), IgM (κ), free kappa light chains, and Bence-Jones kappa light chains. It is non-reactive with lambda light chains.

Application

Monoclonal Anti-Human Kappa Light Chain (Bound and Free) antibody produced in mouse has been used for:
  • immunodetection
  • in western blot analysis
  • in intracellular kappa light chain staining for flow cytometric analysis

Physical form

Monoclonal Anti-Human Kappa Light Chain (Bound and Free) antibody produced in mouse is provided as ascites fluid with 0.1% sodium azide (see MSDS) as a preservative.

Storage and Stability

For continuous use, store at 2-8 °C. For extendedstorage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Upregulated expression of kappa light chain by Epstein-Barr virus encoded latent membrane protein 1 in nasopharyngeal carcinoma cells via NF-kappaB and AP-1 pathways
Liu H, et al.
Cellular Signalling, 19(2), 419-427 (2007)
Chloe L Stoyle et al.
The Biochemical journal, 474(18), 3179-3188 (2017-08-09)
Rodent monoclonal antibodies with specificity towards important biological targets are developed for therapeutic use by a process of humanisation. This process involves the creation of molecules, which retain the specificity of the rodent antibody but contain predominantly human coding sequence.
Heterogeneity of aberrant immunoglobulin expression in cancer cells
Hu D, et al.
Cellular & Molecular Immunology, 8(6), 479-479 (2011)
Chromosomal location of human kappa and lambda immunoglobulin light chain constant region genes
McBRIDE OW, et al.
The Journal of Experimental Medicine, 155(5), 1480-1490 (1982)
Expression and secretion of immunoglobulin alpha heavy chain with diverse VDJ recombinations by human epithelial cancer cells
Zheng H, et al.
Molecular Immunology, 44(9), 2221-2227 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service