Skip to Content
Merck
All Photos(1)

Key Documents

K1385

Sigma-Aldrich

KN-93

≥98% (HPLC)

Synonym(s):

N-[2-[N-(4-Chlorocinnamyl)-N-methylaminomethyl]phenyl]-N-(2-hydroxyethyl)-4-methoxybenzenesulfonamide phosphate salt, N-[2-[[[3-(4′-Chlorophenyl)-2-propenyl]methylamino]methyl]phenyl]-N-(2-hydroxyethyl)-4′-methoxybenzenesulfonamide phosphate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H29ClN2O4S · H3PO4
CAS Number:
Molecular Weight:
599.03
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: 1 mg/mL
H2O: soluble

storage temp.

2-8°C

SMILES string

OP(O)(O)=O.COc1ccc(cc1)S(=O)(=O)N(CCO)c2ccccc2CN(C)C\C=C\c3ccc(Cl)cc3

InChI

1S/C26H29ClN2O4S.H3O4P/c1-28(17-5-6-21-9-11-23(27)12-10-21)20-22-7-3-4-8-26(22)29(18-19-30)34(31,32)25-15-13-24(33-2)14-16-25;1-5(2,3)4/h3-16,30H,17-20H2,1-2H3;(H3,1,2,3,4)/b6-5+;

InChI key

NNKJTPOXLIILMB-IPZCTEOASA-N

Looking for similar products? Visit Product Comparison Guide

Application

KN-93 has been used:
  • as an inhibitor to block αCaMKII (Ca2+/calmodulin-dependent protein kinase IIα)
  • as CaMKII inhibitor to pre-treat hippocampal slices
  • to treat the cells, to alter the cytoskeleton (CSK) structure and function during the experiment

Biochem/physiol Actions

KN-93 is a selective Ca2+/calmodulin-dependent protein kinase II inhibitor, which has been implicated in the regulation of smooth muscle contractility. CaM kinase II activation was inhibited by KN-93 pretreatment (IC50 ~1 μM). KN-93 inhibited histamine-induced tonic force maintenance, whereas early force development and MLC20 phosphorylation responses during the entire time course were unaffected. Both force development and maintenance in response to KCl were inhibited by KN-93. Rapid increases in KCl-induced MLC20 phosphorylation were also inhibited by KN-93, whereas steady-state MLC20 phosphorylation responses were unaffected. In contrast, phorbol 12,13-dibutyrate (PDBu) did not activate CaM kinase II and PDBu-stimulated force development was unaffected by KN-93. Thus KN-93 appears to target a step(s) essential for force maintenance in response to physiological stimuli, suggesting a role for CaM kinase II in regulating tonic contractile responses in arterial smooth muscle. Pharmacological activation of protein kinase C bypasses the KN-93 sensitive step.

Features and Benefits

This compound is featured on the Ca/CaMKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

Photosensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potentiation of Schaffer-Collateral CA1 Synaptic Transmission by eEF2K and p38 MAPK Mediated Mechanisms
Weng W, et al.
Frontiers in Cellular Neuroscience, 247-247 (2016)
Thomas Hennig et al.
PLoS pathogens, 14(3), e1006954-e1006954 (2018-03-27)
Lytic herpes simplex virus 1 (HSV-1) infection triggers disruption of transcription termination (DoTT) of most cellular genes, resulting in extensive intergenic transcription. Similarly, cellular stress responses lead to gene-specific transcription downstream of genes (DoG). In this study, we performed a
Wingless-type mammary tumor virus integration site family, member 5A (Wnt5a) regulates human immunodeficiency virus type 1 (HIV-1) envelope glycoprotein 120 (gp120)-induced expression of pro-inflammatory cytokines via the Ca2+/calmodulin-dependent protein kinase II (CaMKII) and c-Jun N-terminal kinase (JNK) signaling pathways
Li B, et al.
Test, 288(19), 13610-13619 (2013)
Kristen H Jardine et al.
Scientific reports, 10(1), 9209-9209 (2020-06-10)
Reactivated long-term memories can become labile and sensitive to modification. Memories in this destabilized state can be weakened or strengthened, but there is limited research characterizing the mechanisms underlying retrieval-induced qualitative updates (i.e., information integration). We have previously implicated cholinergic
Differential Responses of Cultured MC3T3-E1 Cells to Dynamic and Static Stimulated Effect of Microgravity in Cell Morphology, Cytoskeleton Structure and Ca2+ Signaling
Luo M, et al.
mBio, 13(2), 137-157 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service