Skip to Content
Merck
All Photos(2)

Key Documents

I5523

Sigma-Aldrich

Insulin from porcine pancreas

powder, ≥27 USP units/mg

Synonym(s):

Insulin from hog pancreas

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C256H381N65O76S6
CAS Number:
Molecular Weight:
5777.54
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32
biological source:
Porcine pancreas
form:
powder

biological source

Porcine pancreas

sterility

non-sterile

Assay

≥98%

form

powder

specific activity

≥27 USP units/mg

technique(s)

cell culture | mammalian: suitable

solubility

0.01 M HCl: 5 mg/mL, clear, colorless

cation traces

Zn: 0.20-0.70%

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

InChI key

NOESYZHRGYRDHS-ZYCCASTOSA-N

Gene Information

pig ... INS(397415)

Looking for similar products? Visit Product Comparison Guide

Application

Insulin from porcine pancreas has been used:
  • for culturing of the primary tumors obtained from mice and porcine preantral follicles
  • to characterize hormones involved in postnatal chicken preadipocyte differentiation in vitro
  • to check the reduction ability of the Escherichia coli thiol disulfide exchange proteins (DsbA, DsbC, DsbG) towards insulin

Biochem/physiol Actions

Regulates the cellular uptake, utilization, and storage of glucose, amino acids, and fatty acids and inhibits the breakdown of glycogen, protein, and fat.
Two-chain polypeptide hormone produced by the β-cells of pancreatic islets. Its molecular weight is ~5800 Da. The α and β chains are joined by two interchain disulfide bonds. The α chain contains an intrachain disulfide bond. Insulin regulates the cellular uptake, utilization, and storage of glucose, amino acids, and fatty acids and inhibits the breakdown of glycogen, protein, and fat.

Other Notes

InChI: 1S/C256H381N65O76S6/c1-29-131(23)205(313-193(337)104-258)251(391)317-204(130(21)22)247(387)288-159(75-82-200(347)348)217(357)282-156(71-78-189(262)333)221(361)308-183-116-401-402-117-184-242(382)305-178(111-323)239(379)294-162(88-123(7)8)224(364)295-168(95-140-53-61-146(327)62-54-140)227(367)283-154(69-76-187(260)331)218(358)290-161(87-122(5)6)222(362)285-158(74-81-199(345)346)220(360)302-174(101-190(263)334)234(374)298-170(97-142-57-65-148(329)66-58-142)230(370)309-182(241(381)304-176(255(396)397)103-192(265)336)115-400-399-114-181(213(353)272-107-194(338)278-153(72-79-197(341)342)216(356)281-151(51-42-84-270-256(266)267)211(351)271-108-195(339)279-166(93-138-46-36-32-37-47-138)226(366)297-167(94-139-48-38-33-39-49-139)229(369)299-171(98-143-59-67-149(330)68-60-143)237(377)320-208(136(28)326)253(393)321-85-43-52-186(321)245(385)286-152(50-40-41-83-257)214(354)277-134(26)254(394)395)311-249(389)203(129(19)20)316-235(375)164(90-125(11)12)292-228(368)169(96-141-55-63-147(328)64-56-141)296-223(363)160(86-121(3)4)289-209(349)133(25)276-215(355)157(73-80-198(343)344)287-246(386)202(128(17)18)315-236(376)165(91-126(13)14)293-232(372)173(100-145-106-269-120-275-145)301-238(378)177(110-322)280-196(340)109-273-212(352)180(113-398-403-118-185(310-243(183)383)244(384)319-207(135(27)325)252(392)306-179(112-324)240(380)318-206(132(24)30-2)250(390)312-184)307-225(365)163(89-124(9)10)291-231(371)172(99-144-105-268-119-274-144)300-219(359)155(70-77-188(261)332)284-233(373)175(102-191(264)335)303-248(388)201(127(15)16)314-210(350)150(259)92-137-44-34-31-35-45-137/h31-39,44-49,53-68,105-106,119-136,150-186,201-208,322-330H,29-30,40-43,50-52,69-104,107-118,257-259H2,1-28H3,(H2,260,331)(H2,261,332)(H2,262,333)(H2,263,334)(H2,264,335)(H2,265,336)(H,268,274)(H,269,275)(H,271,351)(H,272,353)(H,273,352)(H,276,355)(H,277,354)(H,278,338)(H,279,339)(H,280,340)(H,281,356)(H,282,357)(H,283,367)(H,284,373)(H,285,362)(H,286,385)(H,287,386)(H,288,387)(H,289,349)(H,290,358)(H,291,371)(H,292,368)(H,293,372)(H,294,379)(H,295,364)(H,296,363)(H,297,366)(H,298,374)(H,299,369)(H,300,359)(H,301,378)(H,302,360)(H,303,388)(H,304,381)(H,305,382)(H,306,392)(H,307,365)(H,308,361)(H,309,370)(H,310,383)(H,311,389)(H,312,390)(H,313,337)(H,314,350)(H,315,376)(H,316,375)(H,317,391)(H,318,380)(H,319,384)(H,320,377)(H,341,342)(H,343,344)(H,345,346)(H,347,348)(H,394,395)(H,396,397)(H4,266,267,270)/t131-,132-,133-,134-,135+,136+,150-,151-,152-,153-,154-,155-,156-,157-,158-,159-,160-,161-,162-,163-,164-,165-,166-,167-,168-,169-,170-,171-,172-,173-,174-,175-,176-,177-,178-,179-,180-,181-,182-,183-,184-,185-,186-,201-,202-,203-,204-,205-,206-,207-,208-/m0/s1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eliana Mariño et al.
Diabetes, 57(2), 395-404 (2007-11-21)
B-cells are important for disease pathogenesis in the nonobese diabetic (NOD) mouse model of type 1 diabetes. Recent studies demonstrate that marginal-zone B-cells (MZBs), which connect innate with adaptive immune responses, are increased in NOD mice. However, beyond this, the
M T Ackermans et al.
Annals of clinical biochemistry, 45(Pt 4), 429-430 (2008-06-28)
The aim of this study is to compare the performance of three commonly used insulin assays with respect to the detection of exogenous human and porcine insulin added to human or rat plasma. The DPC Immulite human insulin assay, the
P H Bessette et al.
The Journal of biological chemistry, 274(12), 7784-7792 (1999-03-13)
We have characterized in vivo and in vitro the recently identified DsbG from Escherichia coli. In addition to sharing sequence homology with the thiol disulfide exchange protein DsbC, DsbG likewise was shown to form a stable periplasmic dimer, and it
Dominik Pesta et al.
Frontiers in endocrinology, 12, 693683-693683 (2021-10-19)
The rs540467 SNP in the NDUFB6 gene, encoding a mitochondrial complex I subunit, has been shown to modulate adaptations to exercise training. Interaction effects with diabetes mellitus remain unclear. We assessed associations of habitual physical activity (PA) levels with metabolic
Michal M Masternak et al.
The journals of gerontology. Series A, Biological sciences and medical sciences, 65(1), 24-30 (2009-11-13)
The disruption of the growth hormone (GH) axis in mice promotes insulin sensitivity and is strongly correlated with extended longevity. Ames dwarf (Prop1(df), df/df) mice are GH, prolactin (PRL), and thyrotropin (TSH) deficient and live approximately 50% longer than their

Protocols

Chromatographic technique identifies insulin variants efficiently, aiding in pharmacokinetics.

Chromatographic technique identifies insulin variants efficiently, aiding in pharmacokinetics.

Chromatographic technique identifies insulin variants efficiently, aiding in pharmacokinetics.

Chromatographic technique identifies insulin variants efficiently, aiding in pharmacokinetics.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service