Skip to Content
Merck
All Photos(1)

Key Documents

I5278

Sigma-Aldrich

Insulin-Like Growth Factor Binding Protein-3 human

recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture, ≥95% (SDS-PAGE)

Synonym(s):

IGF Binding Protein-3, IGFBP-3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

Quality Level

recombinant

expressed in NSO cells

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

0.01-0.25 μg/mL

mol wt

29 kDa

packaging

pkg of 25 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IGFBP3(3486)

Related Categories

Biochem/physiol Actions

Human IGFBP-3 is the major insulin-like growth factor (IGF) binding protein in plasma, where it exists in a ternary complex with IGF-I or IGF-II and the acid-labile subunit (ALS).
IGFBPs (insulin like growth factor binding proteins) regulate bioavailability of IGFs (insulin like growth factors) in cells. Binding between IGFs and IGFBPs reduces the availability of the IGFs. However, IGFBPs also concentrate IGFs in specific areas to potentiate their effects at right time.[1] IGFBP3 binds to IGF-1.[2] It is a tumor suppressor and is downregulated in many cancers.[3] In serum, IGFs, IGFBP3 and the acid labile subunit (ALS) exist together as a complex. This complex is important for increasing the half life of serum IGFs.[4]

Physical form

Lyophilized from a 0.2 μm filtered solution in 30% acetonitrile and 0.1% TFA.

Analysis Note

The biological acitivity is measured as the ability to inhibit the biological activity of 14 ng/ml recombinant human IGF-I on MCF-7 cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Sun Kim et al.
Molecular and cellular biochemistry, 403(1-2), 107-113 (2015-02-11)
Nuclear factor-kappaB (NF-κB) is a transcription factor that is activated in various neoplasms, including gastric cancer. Insulin-like growth factor binding protein-3 (IGFBP-3) is a potent tumor suppressor and is significantly suppressed in a variety of cancers. Although IGFBP-3 has been
Kelley, K.M., et al.
The International Journal of Biochemistry & Cell Biology, 28, 619-637 (1995)
Chuan-Wei Yang et al.
Journal of the American Medical Directors Association, 16(5), 365-370 (2014-12-31)
Previous studies have demonstrated the polymorphisms of insulinlike growth factor-1 (IGF-1) and its binding protein-3 (IGFBP3) genes could affect the circulating IGF-1 level. Moreover, the serum IGF-1 level was correlated with muscle size. This study aimed to explore the effect
Shoshana Yakar et al.
The Journal of clinical investigation, 110(6), 771-781 (2002-09-18)
IGF-1 is a growth-promoting polypeptide that is essential for normal growth and development. In serum, the majority of the IGFs exist in a 150-kDa complex including the IGF molecule, IGF binding protein 3 (IGFBP-3), and the acid labile subunit (ALS).
M L Cubbage et al.
The Journal of biological chemistry, 265(21), 12642-12649 (1990-07-25)
Insulin-like growth factor binding protein-3 (IGFBP-3) can modulate the mitogenic and metabolic effects of the insulin-like growth factors (IGFs). IGFBP-3 protein levels are developmentally regulated and influenced by a number of hormonal stimuli both in vitro and in vivo. As

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service