Skip to Content
Merck
All Photos(1)

Key Documents

I3786

Sigma-Aldrich

Protease Inhibitor Cocktail

Animal-Free, DMSO solution, for the inhibition of serine, cysteine, aspartic proteases and aminopeptidases, for use with mammalian cell and tissue extract, DMSO solution

Synonym(s):

Protease inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77
form:
DMSO solution
solubility:
water: soluble
storage temp.:
−20°C

Product Name

Protease Inhibitor Cocktail, Animal Component Free, for use with mammalian cell and tissue extract, DMSO solution

Quality Level

form

DMSO solution

solubility

water: soluble

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Components of this cocktail are of non-animal origin and no animal products were used in the component production process. All vessels and instruments used for the cocktail production are dedicated for animal component free production and have never encountered an animal product. Aprotinin used in this cocktail is a recombinant bovine protein expressed in plants (Nicotiana).
This protease inhibitor cocktail has been optimized and tested for mammalian cell and tissue extracts.

Specificity

Inhibits serine, cysteine, aspartic proteases, and aminopeptidases.

Application

Animal component free (ACF). Optimized and tested for general use. Can also be used for purification of histidine-tagged proteins, as chelating agents have been omitted.

Components

AEBSF
Aprotinin
Bestatin
E-64
Leupeptin
Pepstatin A

Physical form

The components in this ACF protease inhibitor cocktail do not originate from animal sources and can thus be used in the production of animal-free proteins.
This ACF cocktail is supplied as a ready-to-use solution in DMSO. One ml of the cocktail solution is recommended for the inhibition of endogenous enzymes found in 100 mL of lysate from 20 g (wet weight) of bovine liver, or 10 mL of cell lysate obtained from CHO cells at a cell density of 108 cells per mL.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yue Gao et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 129, 110396-110396 (2020-06-25)
Systemic lupus erythematosus (SLE) is an autoimmune disease that is mainly caused by excessive accumulation of autoantibodies that target autoantibodies such as nucleic acids. T helper (Th) cell have been associated with the development of SLE. Typically, different subsets of
Paulo Emílio Corrêa Leite et al.
Toxicology in vitro : an international journal published in association with BIBRA, 29(5), 819-827 (2015-03-21)
Gold nanoparticles (AuNP) have been widely used for many applications, including as biological carriers. A better understanding concerning AuNP safety on muscle cells is crucial, since it could be a potential tool in the nanomedicine field. Here, we describe the
Wimonrat Panpetch et al.
Frontiers in immunology, 11, 561652-561652 (2020-10-27)
Obesity induces gut leakage and elevates serum lipopolysaccharide (LPS), a major cell wall component of Gram-negative bacteria, through gut translocation. Because Candida albicans is prominent in human gut but not in mouse, C. albicans, a source of (1→3)-β-D-glucan (BG) in
Mo Shen et al.
Molecular medicine reports, 16(1), 937-942 (2017-06-07)
The role of inflammation in tumorigenesis and development is currently well established. Lymphotoxin β receptor (LTβR) activation induces canonical and noncanonical nuclear factor (NF)‑κB signaling pathways, which are linked to inflammation‑induced carcinogenesis. In the present study, 5,637 bladder cancer cells were
Douglas Esteves Teixeira et al.
Biochimica et biophysica acta. Molecular basis of disease, 1866(11), 165907-165907 (2020-08-02)
High salt diet (HSD), considered a public health problem worldwide, is associated with chronic degenerative diseases including renal diseases. However, little is known about the effects of HSD on renal function independently of the development of hypertension. To address the

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service