Skip to Content
Merck
All Photos(1)

Key Documents

I1637

Sigma-Aldrich

Ipratropium bromide monohydrate

≥98% (TLC), powder

Synonym(s):

Atropine isopropyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30BrNO3 · H2O
CAS Number:
Molecular Weight:
430.38
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (TLC)

form

powder

color

white

solubility

H2O: 10 mg/mL

originator

Boehringer Ingelheim

SMILES string

O.[Br-].CC(C)[N@+]1(C)[C@H]2CC[C@@H]1C[C@@H](C2)OC(=O)C(CO)c3ccccc3

InChI

1S/C20H30NO3.BrH.H2O/c1-14(2)21(3)16-9-10-17(21)12-18(11-16)24-20(23)19(13-22)15-7-5-4-6-8-15;;/h4-8,14,16-19,22H,9-13H2,1-3H3;1H;1H2/q+1;;/p-1/t16-,17+,18+,19?,21?;;

InChI key

KEWHKYJURDBRMN-XSAPEOHZSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Ipratropium bromide monohydrate has been used for the inhibition of peripheral muscarinic receptors in rats.

Biochem/physiol Actions

Ipratropium bromide monohydrate is a nonselective muscarinic acetylcholine receptor antagonist and a bronchodilator. In nebulized form, it is used for treating asthma and obstructive pulmonary diseases. It acts as a β2 adrenoreceptor agonist. Ipratropium bromide monohydrate is an anticholinergic agent and competes with acetylcholine. It promotes bronchodilation by stimulating sympathetic pathways.

Features and Benefits

This compound was developed by Boehringer Ingelheim. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Other Notes

N-isopropyl quaternary salt of atropine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

For COPD a combination of ipratropium bromide and albuterol sulfate is more effective than albuterol base.
Campbell S
Archives of Internal Medicine, 159(2), 156-160 (1999)
Bilateral acute angle closure developing due to use of ipratropium bromide and salbutamol.
Kola M, et al.
International Ophthalmology, 38(1), 385-388 (2018)
E B Haddad et al.
British journal of pharmacology, 127(2), 413-420 (1999-06-29)
1. In this study we have evaluated the pharmacological profile of the muscarinic antagonist glycopyrrolate in guinea-pig and human airways in comparison with the commonly used antagonist ipratropium bromide. 2. Glycopyrrolate and ipratropium bromide inhibited EFS-induced contraction of guinea-pig trachea
Comparative characterization of lung muscarinic receptor binding after intratracheal administration of tiotropium, ipratropium, and glycopyrrolate.
Ogoda M, et al.
Journal of Pharmacological Sciences, 115(3), 374-382 (2011)
H Groeben et al.
Anesthesiology, 85(4), 867-873 (1996-10-01)
Two major groups of drugs are available to prevent bronchoconstriction: beta-agonists and muscarinic blocking agents. Ipratropium is the most commonly used anticholinergic agent to treat chronic obstructive pulmonary disease. The authors studied anti-muscarinic agents to determine if they are as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service