Skip to Content
Merck
All Photos(3)

Key Documents

I1406

Sigma-Aldrich

Irinotecan hydrochloride

powder, ≥97% (HPLC)

Synonym(s):

(S)-4,11-diethyl-3,4,12,14-tetrahydro-4-hydroxy-3,14-dioxo-1H-pyrano[3′,4′:6,7]indolizino[1,2-b]quinolin-9-yl ester, CPT-11, [1,4′-Bipiperidine]-1′-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H38N4O6 · HCl
CAS Number:
Molecular Weight:
623.14
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Irinotecan hydrochloride, topoisomerase inhibitor

biological source

plant (Fructus camptothecae)

Assay

≥97% (HPLC)

form

powder

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

Cl.CCc1c2CN3C(=O)C4=C(C=C3c2nc5ccc(OC(=O)N6CCC(CC6)N7CCCCC7)cc15)[C@@](O)(CC)C(=O)OC4

InChI

1S/C33H38N4O6.ClH/c1-3-22-23-16-21(43-32(40)36-14-10-20(11-15-36)35-12-6-5-7-13-35)8-9-27(23)34-29-24(22)18-37-28(29)17-26-25(30(37)38)19-42-31(39)33(26,41)4-2;/h8-9,16-17,20,41H,3-7,10-15,18-19H2,1-2H3;1H/t33-;/m0./s1

InChI key

GURKHSYORGJETM-WAQYZQTGSA-N

Gene Information

human ... TOP1(7150)

Application

Irinotecan hydrochloride has been used:
  • in combination with 5-fluorouracil for screening growth inhibitory functionality in MDA-MB-231 breast cancer cells.[1]
  • in chemosensitivity screening of high-grade appendiceal (HGA) and low-grade appendiceal (LGA) organoids.[2]
  • as a chemotherapeutic agent in the cytotoxicity studies in combination with heat shock proteins inhibitors (HPSC1) in HT29 colon cancer cells.[3]

Biochem/physiol Actions

The anticancer agent, irinotecan, is a prodrug that is converted by tissue carboxylesterase to 7-ethyl-10-hydroxycamptothecin (SN-38), a potent inhibitor of DNA topoisomerase I. Its action is terminated by glucuronidation by UDP glucuronosyl transferase 1A1 (UGT1A1).
The anticancer agent, irinotecan, is a prodrug that is converted by tissue carboxylesterase to 7-ethyl-10-hydroxycamptothecin (SN-38), a potent inhibitor of DNA topoisomerase I. Its action is terminated by glucuronidation by UDP glucuronosyl transferase 1A1 (UGT1A1). It proves useful in radiation treatment of tumors by sensitizing tissue to radiation damage.
The anticancer agent, irinotecan, is a prodrug that is converted by tissue carboxylesterase to 7-ethyl-10-hydroxycamptothecin (SN-38), a potent inhibitor of DNA topoisomerase I. Its action is terminated by glucuronidation by UDP glucuronosyl transferase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesco Crea et al.
Molecular cancer therapeutics, 8(7), 1964-1973 (2009-06-18)
Irinotecan is a topoisomerase-I (Top-I) inhibitor used for the treatment of colorectal cancer. DNA demethylating agents, including 5-azacytidine (5-aza), display synergistic antitumor activity with several chemotherapy drugs. 5-Aza may enhance irinotecan cytotoxicity by at least one of the following mechanisms:
Re-examining HSPC1 inhibitors
Lee S, et al.
Cell Stress & Chaperones, 22(2), 293-306 (2017)
Appendiceal Cancer Patient-Specific Tumor Organoid Model for Predicting Chemotherapy Efficacy Prior to Initiation of Treatment: A Feasibility Study
Votanopoulos K, et al.
Annals of Surgical Oncology, 26(1), 139-147 (2019)
T C Chang et al.
Cancer chemotherapy and pharmacology, 75(3), 579-586 (2015-01-13)
To define the dose-limiting toxicity (DLT), maximum tolerated dose (MTD) and pharmacokinetics (PK) of PEP02, a novel liposome-encapsulated irinotecan, in patients with advanced refractory solid tumors. Patients were enrolled in cohorts of one to three to receive escalating dose of
Olivia Jones et al.
Scientific reports, 11(1), 8967-8967 (2021-04-28)
Cholangiocarcinoma (CCA) is a rare biliary tract cancer with a low five-year survival rate and high recurrence rate after surgical resection. Currently treatment approaches include systemic chemotherapeutics such as FOLFIRINOX, a chemotherapy regimen is a possible treatment for severe CCA

Articles

Explore how antibiotics, particularly quinolones, inhibit nucleic acid synthesis by targeting topoisomerases, crucial for DNA replication processes.

Explore how antibiotics, particularly quinolones, inhibit nucleic acid synthesis by targeting topoisomerases, crucial for DNA replication processes.

Explore how antibiotics, particularly quinolones, inhibit nucleic acid synthesis by targeting topoisomerases, crucial for DNA replication processes.

Explore how antibiotics, particularly quinolones, inhibit nucleic acid synthesis by targeting topoisomerases, crucial for DNA replication processes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service