Skip to Content
Merck
All Photos(1)

Key Documents

I1142

Sigma-Aldrich

Monoclonal Anti-Interferon-γ antibody produced in rat

clone 37895, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-IFN-γ

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

37895, monoclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

flow cytometry: suitable
neutralization: suitable
western blot: suitable

isotype

IgG2a

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Ifng(15978)

Related Categories

General description

Interferon-γ (IFN-γ) belongs to Type II class of interferons and is produced by T helper cells (specifically, Th1 cells), natural killer (NK) and natural killer T (NKT) cells and cytotoxic T cells (TC cells) . It has various biological functions including antiviral activity like inhibition of cell or tumor growth, act as a potent modulator of immune responses and modifies cellular processes as well as stimulates the differentiation of B cells into immunoglobulin-producing cells. Monoclonal Anti-Interferon-γ antibody can be used in immunoblotting. Rat anti-Interferon-γ antibody has shown no cross-reactivity for recombinant human IFN-γ or recombinant rat IFN-γ.

Specificity

The antibody does not cross-react with recombinat human IFN-γ or recombinant rat IFN-γ.

Immunogen

recombinant mouse IFN-γ.

Application

Monoclonal Anti-Interferon-γ antibody can be used in neutralization and indirect ELISA.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline and 5% trehalose.

Preparation Note

Purified from cell culture supernatant using protein G.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Gresser et al.
Proceedings of the National Academy of Sciences of the United States of America, 66(4), 1052-1058 (1970-08-01)
Mouse interferon preparations inhibited the multiplication of mouse leukemia L 1210 cells in stationary suspension cultures. The degree of inhibition was found to correlate with the antiviral titer of the interferon preparations. The factor(s) responsible for inhibition of L 1210
P B Fisher et al.
Pharmacology & therapeutics, 27(2), 143-166 (1985-01-01)
As described in this review, both partially purified and recombinant interferons are potent modulators of differentiation in diverse cell culture systems (Table 2). Depending on the target cell, interferon exerts either an inhibitory or an inductive effect on cell differentiation.
J Vilcek et al.
Journal of immunology (Baltimore, Md. : 1950), 135(3), 1851-1856 (1985-09-01)
Interferon (IFN)-gamma production, stimulated by the addition of exogenous interleukin (IL) 2, T cell mitogens, or tuberculin purified protein derivative (PPD) was studied in cultures of separated human mononuclear cells or unseparated peripheral blood leukocytes (PBL). IFN-gamma was induced by
Jamie R Schoenborn et al.
Advances in immunology, 96, 41-101 (2007-11-06)
Interferon-gamma (IFN-gamma) is crucial for immunity against intracellular pathogens and for tumor control. However, aberrant IFN-gamma expression has been associated with a number of autoinflammatory and autoimmune diseases. This cytokine is produced predominantly by natural killer (NK) and natural killer
P D Benech et al.
The Journal of experimental medicine, 176(4), 1115-1123 (1992-10-01)
The human Fc gamma RI (CD64) is a high affinity receptor for the Fc portion of immunoglobulin (Ig), and its constitutively low expression on the cell surface of monocyte/macrophage and neutrophils is selectively upregulated by interferon gamma (IFN-gamma) treatment (Perussia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service