Skip to Content
Merck
All Photos(1)

Documents

I0531

Sigma-Aldrich

Interleukin-18 from rat

>97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

rat

Quality Level

recombinant

expressed in E. coli

Assay

>97% (SDS-PAGE)

form

lyophilized powder

potency

10-60 ng/mL ED50

mol wt

predicted mol wt ~18.4 kDa

packaging

pkg of 25 μg

impurities

endotoxin, tested

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

rat ... Il18(29197)

Biochem/physiol Actions

Interleukin 18, also known as interferon-γ-inducing factor (IGIF) and IL-1γ, is a proinflammatory cytokine. It is an inducer of IFN-γ and Th1 (T cell helper type 1) responses (cell differentiation, cell-mediated cytotoxicity and inflammation). IL-18 induces the migration of microvascular endothelial cells.

Physical form

Lyophilized from 20 mM Tris , 500 mM NaCl, 0.5 mM EDTA, 1 mM TCEP, 0.05% (w/v) PEG8000 pH 8.0 with 5% Trehalose,and 50 μg BSA per μg IL-18

Analysis Note

The biological activity is measured by its ability to induce mouse IFN-γ production by activated mouse T cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weiyi He et al.
Cytokine, 68(1), 32-39 (2014-05-03)
CD4(+) T cell polarization plays a critical role in the pathogenesis of allergy. How to modulate the skewed CD4(+) T cell polarization is less clear. The specific immunotherapy (SIT) is the only specific remedy for the treatment of allergic diseases;
Jochem W van der Veen et al.
Regulatory toxicology and pharmacology : RTP, 69(3), 371-379 (2014-05-13)
The currently available animal-free methods for the detection of skin sensitizing potential of chemicals seem promising. However, no single method is able to comprehensively represent the complexity of the processes involved in skin sensitization. To ensure a mechanistic basis and
M Dagenais et al.
Mucosal immunology, 9(1), 146-158 (2015-06-04)
Cellular inhibitors of apoptosis proteins (cIAPs) are critical arbiters of cell death and key mediators of inflammation and innate immunity. cIAP2 is frequently overexpressed in colorectal cancer and in regenerating crypts of ulcerative colitis patients. However, its corresponding functions in
Wei Guo et al.
Molecular pain, 10, 35-35 (2014-06-11)
It has been recently recognized that the descending serotonin (5-HT) system from the rostral ventromedial medulla (RVM) in the brainstem and the 5-HT3 receptor subtype in the spinal dorsal horn are involved in enhanced descending pain facilitation after tissue and
Anders Elm Pedersen et al.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica, 123(7), 547-555 (2015-04-25)
TL1A is a TNF-like cytokine which has been shown to co-stimulate TH1 and TH17 responses during chronic inflammation. The expression of this novel cytokine has been investigated in inflammatory disorders like rheumatoid arthritis and inflammatory bowel disease, but little is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service