Skip to Content
Merck
All Photos(1)

Documents

HT25A

Sigma-Aldrich

Elastic Stain

Synonym(s):

Modified Verhoeff Van Gieson Elastic Stain Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121
NACRES:
NA.47

form

solution

Quality Level

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

application(s)

hematology
histology

storage temp.

room temp

Diagnostic Implications

Demonstration of elastic tissue is useful in cases of emphysema (atrophy of elastic tissue), arteriosclerosis (thinning and loss of elastic fibers), and various other vascular diseases.

Kit Components Only

Product No.
Description

  • Ferric Chloride (kit only) 125 mL

  • Hematoxylin, Alcoholic (kit only) 2 x 200

  • Weigert's Iodine (kit only) 180 mL

Kit Components Also Available Separately

Product No.
Description
SDS

  • HT254Van Gieson Solution , for in vitro diagnostic use 250 mLSDS

Signal Word

Danger

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 2

Target Organs

Eyes,Central nervous system, Thyroid

Storage Class Code

3 - Flammable liquids

Flash Point(F)

55.9 °F - closed cup

Flash Point(C)

13.3 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dimitria Bonizol Camasão et al.
Biomaterials science, 8(12), 3536-3548 (2020-06-02)
Natural polymers are commonly used as scaffolds for vascular tissue engineering. The recognized biological properties of this class of materials are often counterbalanced by their low mechanical performance. In this work, recombinant elastin-like polypeptides (or elastin-like recombinamers, ELRs) were mixed
Katrine E Karmisholt et al.
Lasers in surgery and medicine, 49(2), 189-197 (2016-11-20)
Caesarean section (c-section) scars can be pose functional and cosmetic challenges and ablative fractional laser (AFXL) treatment may offer benefit to patients. We evaluated textural and color changes over time in AFXL-treated versus untreated control scars. A randomized, controlled, intra-individual
Peng Zhu et al.
The Journal of surgical research, 200(2), 707-713 (2015-09-17)
Allograft arteriopathy is still a leading cause of late organ failure. The aortic allograft model in mice has been used to study chronic rejection and has given useful information in the development of graft arteriosclerosis. However, the technical difficulties of
Xiao-Fang Huang et al.
Acta pharmacologica Sinica, 40(2), 192-198 (2018-05-20)
Abdominal aortic aneurysms (AAAs) are a chronic vascular disease characterized by pathological luminal dilation. Aortic rupture is the fatal consequence of AAAs. Ginkgo biloba extracts (GBEs), a natural herb extract widely used as food supplements, drugs, and cosmetics, has been
Basant Dhital et al.
Archives of biochemistry and biophysics, 666, 127-137 (2019-03-28)
Female reproductive tissues undergo significant alterations during pregnancy, which may compromise the structural integrity of extracellular matrix proteins. Here, we report on modifications of elastic fibers, which are primarily composed of elastin and believed to provide a scaffold to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service