Skip to Content
Merck
All Photos(3)

Documents

HPA046259

Sigma-Aldrich

Anti-AQP8 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Aquaporin 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500- 1:1000

immunogen sequence

EIAMCEPEFGNDKAREPSVGGRWRVSWYERF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AQP8(343)

General description

Aquaporin 8 (AQP8) belongs to the water channel proteins, aquaporins (AQPs). The protein is expressed in human pancreas and colon. It encodes a 261 amino acid protein. The gene is located on human chromosome 16p12.1.

Immunogen

aquaporin 8 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Aquaporin 8 (AQP8) regulates water metabolism. It contributes to pathogenesis of inflammatory disease. The protein facilitates the bidirectional transport of water and hydrogen peroxide across biological membranes. Hepatocyte mitochondrial aquaporin-8 (mtAQP8) functions as a multifunctional membrane channel protein, which enables the uptake of ammonia for its detoxification to urea and the mitochondrial release of hydrogen peroxide. Knockdown of mitochondrial AQP8 is linked to necrotic cell death in human neoplastic hepatic cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST81836

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cholesterol can modulate mitochondrial aquaporin-8 expression in human hepatic cells
Danielli M, et al.
IUBMB Life, 69(5), 341-346 (2017)
Evidence for necrosis, but not apoptosis, in human hepatoma cells with knockdown of mitochondrial aquaporin-8
Marchissio MJ, et al.
Apoptosis, 19(5), 851-859 (2014)
Cloning and functional expression of human aquaporin8 cDNA and analysis of its gene
Koyama N, et al.
Genomics, 54(1), 169-172 (1998)
Stress regulates aquaporin-8 permeability to impact cell growth and survival
Medrano-Fernandez I, et al.
Antioxidants & Redox Signaling, 24(18), 1031-1044 (2016)
Differential expression of aquaporin 8 in human colonic epithelial cells and colorectal tumors
Fischer H, et al.
BMC Physiology, 1(1), 1-1 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service