Skip to Content
Merck
All Photos(8)

Key Documents

HPA044682

Sigma-Aldrich

Anti-PLIN4 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-KIAA1881, Anti-S3-12, Anti-perilipin 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:2500-1:5000
western blot: 0.04-0.4 μg/mL

immunogen sequence

GVDTSKTVLTGTKDTVCSGVTGAMNVAKGTIQTGVDTTKTVLTGTKNTVCSGVTGAVNLAKEAIQGGLDTTKSMVMGTKDTMSTGLTGAANVAKGAMQTGLNTTQNIATGT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLIN4(729359)

General description

Perilipin 4 (PLIN4) is a lipid droplet protein, encoded by the gene mapped to human chromosome 19q13.3-13.4. PLIN4 is highly expressed in white adipose tissue and is also expressed at low levels in heart and skeletal muscle. The encoded protein and PLIN5 exhibit distinct though overlapping tissue expression pattern.

Immunogen

perilipin 4 recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Perilipin 4 (PLIN4) might be involved in the formation of lipid droplets (LDs) in adipocytes. Perilipins play a vital role in regulation of lipid turnover. In mice, knockdown of this gene downregulates Plin5 expression and decreases cardiac lipid accumulation. Mutation in the gene is linked to development of gastric cancer. In addition, variation in the gene might also lead to obesity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST82778

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inactivation of Plin4 downregulates Plin5 and reduces cardiac lipid accumulation in mice.
Chen W
American Journal of Physiology. Endocrinology and Metabolism, 304(7) (2013)
Whole genome and transcriptome sequencing of matched primary and peritoneal metastatic gastric carcinoma.
Zhang J
Scientific Reports (2015)
Perilipin 4 in human skeletal muscle: localization and effect of physical activity.
Pourteymour S
Physiological Reports, 3 (2015)
Shirin Pourteymour et al.
Physiological reports, 3(8), doi:10-doi:10 (2015-08-13)
Perilipins (PLINs) coat the surface of lipid droplets and are important for the regulation of lipid turnover. Knowledge about the physiological role of the individual PLINs in skeletal muscle is limited although lipid metabolism is very important for muscle contraction.
The PLIN4 variant rs8887 modulates obesity related phenotypes in humans through creation of a novel miR-522 seed site.
Richardson K
PLoS ONE, 6 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service