Skip to Content
Merck
All Photos(3)

Key Documents

HPA043014

Sigma-Aldrich

Anti-RPL19 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DKFZp779D216, Anti-FLJ27452, Anti-L19, Anti-MGC71997, Anti-Ribosomal Protein L19

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
IHC
clone:
polyclonal
species reactivity:
human
citations:
6
technique(s):
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

TNEIANANSRQQIRKLIKDGLIIRKPVTVHSRARCRK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPL19(6143)

General description

Ribosomal protein L19 (RPL19) is a member of L19E super-family of proteins and is a component of the ribosomal large 60S subunit. The gene is located on human chromosome 17q12. Ribosomal protein L19 (RPL19) consists of a promoter between -210 and -250, which is required for gene activity.

Immunogen

ribosomal protein L19 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-RPL19 antibody produced in rabbit has been used in immunofluorescence.

Biochem/physiol Actions

The protein has an extra-protein translational function. Upregulation of RPL19 stimulates michigan cancer foundation-7 (MCF7) breast cancer cells to endoplasmic reticulum (ER) stress-induced cell death. It functions as a tumor antigen in lung adenocarcinoma. The protein is a poor prognostic marker for colorectal cancer. It is an important modulator of cellular phenotype. The protein is used as a prognostic marker for prostate cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST82665

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

siRNA knockdown of ribosomal protein gene RPL19 abrogates the aggressive phenotype of human prostate cancer
Bee A, et al.
PLoS ONE, 6(7), e22672-e22672 (2011)
DNA Ends Alter the Molecular Composition and Localization of Ku Multi-component Complexes
Adelmant G, et al.
Molecular and Cellular Proteomics, mcp-M111 (2012)
Ribosomal protein l19 is a prognostic marker for human prostate cancer
Bee A, et al.
Clinical Cancer Research, 12(7), 2061-2065 (2006)
17q12-21-The pursuit of targeted therapy in breast cancer
Glynn RW, et al.
Cancer Treatment Reviews, 36(3), 224-229 (2010)
The L19 ribosomal protein gene (RPL19): Gene organization, chromosomal mapping, and novel promoter region
Davies B and Fried M
Genomics, 25(2), 372-380 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service