Skip to Content
Merck
All Photos(4)

Key Documents

HPA027052

Sigma-Aldrich

Anti-GPER1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CEPR, Anti-CMKRL2, Anti-DRY12, Anti-FEG-1, Anti-GPCR-Br, Anti-GPER, Anti-GPR30, Anti-LERGU, Anti-LERGU2, Anti-LyGPR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

MDVTSQARGVGLEMYPGTAQPAAPNTTSPELNLSHPLLGTALANGTGELSEHQQYVIGLFLS

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPER(2852)

Immunogen

G protein-coupled estrogen receptor 1 recombinant protein epitope signature tag (PrEST)

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72764

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher A Natale et al.
Cellular and molecular gastroenterology and hepatology, 10(4), 868-880 (2020-05-08)
Female sex is associated with lower incidence and improved clinical outcomes for most cancer types including pancreatic ductal adenocarcinoma (PDAC). The mechanistic basis for this sex difference is unknown. We hypothesized that estrogen signaling may be responsible, despite the fact
Inyoung Lee et al.
Cancer research communications, 3(8), 1594-1606 (2023-08-21)
Despite recent therapeutic advances, the 5-year survival rate for adults with acute myeloid leukemia (AML) is poor and standard-of-care chemotherapy is associated with significant toxicity, highlighting the need for new therapeutic approaches. Recent work from our group and others established
Asako Yamamoto et al.
International journal of molecular sciences, 22(1) (2021-01-02)
As the outermost barrier of the body, skin is a major target of oxidative stress. In the brain, estrogen has been reported synthesized locally and protects neurons from oxidative stress. Here, we explored whether estrogen is also locally synthesized in
Christopher A Natale et al.
eLife, 7 (2018-01-18)
Female sex and history of prior pregnancies are associated with favorable melanoma outcomes. Here, we show that much of the melanoma protective effect likely results from estrogen signaling through the G protein-coupled estrogen receptor (GPER) on melanocytes. Selective GPER activation
Antonio Florido et al.
Biology of sex differences, 13(1), 28-28 (2022-06-12)
Memory consolidation is a process required for the formation of long-term memories. The G-protein-coupled receptor (GPCR) neurokinin-3-receptor (Nk3R) and its interactions with sex hormones seem important for the modulation of fear memory consolidation: Nk3R antagonism in male mice impairs fear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service