Skip to Content
Merck
All Photos(3)

Key Documents

HPA027011

Sigma-Aldrich

Anti-SLC35C2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C20orf5, Anti-CGI-15, Anti-OVCOV1, Anti-bA394O2.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

KALHSRGDGGPKALKGLGSSPDLELLLRSSQREEGDNEEEEYFVAQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC35C2(51006)

General description

Solute carrier family 35 member C2 (SLC35C2) is a nucleotide sugar transporter, which belongs to solute carriers 35 family. It has multiple transmembrane helices and is present on Golgi and endoplasmic reticulum membranes. SLC35C2 is also called OVCOV1 and is mapped to human chromosome 20q13.12. It exists in two isoforms and is majorly expressed in reproductive tissues.

Immunogen

solute carrier family 35 (GDP-fucose transporter), member C2

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Solute carrier family 35 member C2 (SLC35C2) functions to transport sugars from cytosol to organelles. It mediates the transport of guanosine diphosphate (GDP)-fucose and aids fucosylation of epidermal growth factor-like (EGF) repeats in the notch receptors. It also favors notch signalling.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73208

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roles of the nucleotide sugar transporters (SLC35 family) in health and disease
Song Z
Molecular Aspects of Medicine, 34(2-3), 590-600 (2013)
Assignment1 of OVCOV1 (alias CGI-15) to human chromosome 20 band q13. 1? q13. 2 by fluorescent in situ hybridization
Leach R, et al.
Cytogenetic and genome research, 94(3-4), 252-253 (2001)
Identification, molecular characterization, and tissue expression of OVCOV1
Leach R, et al
Mammalian Genome, 13(11), 619-624 (2002)
Small cell ovarian carcinoma: genomic stability and responsiveness to therapeutics
Gamwell L, et al.
Orphanet Journal of Rare Diseases, 8(1), 33-33 (2013)
Slc35c2 promotes Notch1 fucosylation and is required for optimal Notch signaling in mammalian cells
Lu L, et al.
The Journal of Biological Chemistry, 36245?36254-36245?36254 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service