Skip to Content
Merck
All Photos(4)

Documents

HPA024696

Sigma-Aldrich

Anti-FHOD3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-FH1/FH2 domain-containing protein 3, Anti-Formactin-2, Anti-Formin homolog overexpressed in spleen 2, Anti-hFHOS2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

QCGDILTNKRFMLDMLYAHNRKSPDDEEKGDGEAGRTQQEAEAVASLATRISTLQANSQTQDESVRRVDVGCLDNRGSVKAFAEKFNSGDLGRGSISPDAEPNDKVPETAPVQPKTESDYIWDQLM

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FHOD3(80206)

General description

The gene FHOD3 (formin homology 2 domain containing 3) is mapped to human chromosome 18q12. It is expressed in the heart, kidney and brain. The FHOD3 protein belongs to the formin family of proteins. The protein has GTPase-binding, diaphanous inhibitory, formin homologous 1 (FH1), formin homologous 2 (FH2) and diaphanous autoregulation domains.

Immunogen

FH1/FH2 domain-containing protein 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-FHOD3 antibody produced in rabbit has been used in Western blotting. 

Biochem/physiol Actions

FHOD3 (formin homology 2 domain containing 3) is involved in actin polymerization and myofibril formation, thereby helping in sarcomeric organization in cardiomyocytes. Mutations in FHOD3 gene are associated with dilated cardiomyopathy and hypertrophic cardiomyopathy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70045

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J D Cooper et al.
Genes and immunity, 10 Suppl 1, S85-S94 (2009-12-04)
The advent of genome-wide association (GWA) studies has revolutionized the detection of disease loci and provided abundant evidence for previously undetected disease loci that can be pooled together in meta-analysis studies or used to design follow-up studies. A total of
Takuro Arimura et al.
Circulation journal : official journal of the Japanese Circulation Society, 77(12), 2990-2996 (2013-10-04)
Dilated cardiomyopathy (DCM) is characterized by a dilated left ventricular cavity with systolic dysfunction manifested by heart failure. It has been revealed that mutations in genes for cytoskeleton or sarcomere proteins cause DCM. However, the disease-causing mutations can be found
Nikki R Paul et al.
The Journal of cell biology, 210(6), 1013-1031 (2015-09-16)
Invasive migration in 3D extracellular matrix (ECM) is crucial to cancer metastasis, yet little is known of the molecular mechanisms that drive reorganization of the cytoskeleton as cancer cells disseminate in vivo. 2D Rac-driven lamellipodial migration is well understood, but
Eric C Wooten et al.
Circulation. Cardiovascular genetics, 6(1), 10-18 (2012-12-21)
Incomplete penetrance and variable expression of hypertrophic cardiomyopathy (HCM) is well appreciated. Common genetic polymorphisms variants that may affect HCM penetrance and expression have been predicted but are not well established. We performed a case-control genomewide association study to identify

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service