Skip to Content
Merck
All Photos(5)

Key Documents

HPA024616

Sigma-Aldrich

Anti-GREB1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1

Synonym(s):

Greb1 Antibody, Greb1 Antibody - Anti-GREB1 antibody produced in rabbit, Anti-BCTL1, Anti-BTCL1, Anti-KIAA0575, Anti-growth regulation by estrogen in breast cancer 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

EGDIDILLDKFHQENQGHISSSLAASSVTKAASLDVSGTPVCTSYNLEPHSIRPFQLAVAQKLLSHVCSIADSSTQNLDLGSFEKVDFLICIP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GREB1(9687)

General description

The gene GREB1 (growth regulation by estrogen in breast cancer 1) is mapped to human chromosome 2p25.

Immunogen

growth regulation by estrogen in breast cancer 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-GREB1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

GREB1 (growth regulation by estrogen in breast cancer 1) is regulated by estrogen and plays an important role in breast cancer. It controls 17β-estradiol-stimulated breast cancer cell growth. Additionally, it acts as a cofactor for ESR1 (estrogen receptor α)-associated transcription in the cancer cells. It is also linked with ovarian tumor progression. Changes in the expression of it are associated with risk of endometriosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76078

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kendra Hodgkinson et al.
Oncogene, 37(44), 5873-5886 (2018-07-06)
Estrogenic hormone replacement therapy increases the risk of developing ovarian cancer, and estrogen promotes tumour initiation and growth in mouse models of this disease. GREB1 (Growth regulation by estrogen in breast cancer 1) is an ESR1 (estrogen receptor 1)-upregulated protein
Yuechao Zhao et al.
Cancer research, 77(20), 5602-5613 (2017-09-15)
Many estrogen receptor α (ERα)-positive breast cancers develop resistance to endocrine therapy via mutation of ERs whose constitutive activation is associated with shorter patient survival. Because there is now a clinical need for new antiestrogens (AE) against these mutant ERs
Osamu Ichii et al.
The American journal of pathology, 188(9), 2120-2138 (2018-06-24)
The distal tubule (DT) helps regulate blood pressure and electrolytes. We describe a novel, autosomal recessive, morphofunctional DT abnormality in inbred mice evident as columnar alternations and age-related cystic changes. This abnormality developed in both sexes of DBA/2Cr. Similar phenotypes
Sandra Lettlova et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 46(6), 2601-2615 (2018-05-16)
MiRNA-301a-3p is an oncogenic miRNA whose expression is associated with tumor development, metastases and overall poor prognosis. Estrogen receptor α (ERα) is one of the estrogen hormone-activated transcription factors, which regulates a large number of genes and is involved in
Laura A Laviolette et al.
International journal of cancer, 135(5), 1072-1084 (2014-01-29)
Exogenous 17β-estradiol (E2) accelerates the progression of ovarian cancer in the transgenic tgCAG-LS-TAg mouse model of the disease. We hypothesized that E2 has direct effects on ovarian cancer cells and this study was designed to determine the molecular mechanisms by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service