Skip to Content
Merck
All Photos(6)

Key Documents

HPA021451

Sigma-Aldrich

Anti-SLC16A3 antibody produced in rabbit

Anti-SLC16A3 antibody produced in rabbit
1 of 1 reviewers received a sample product or took part in a promotion

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MCT 3, Anti-MCT 4, Anti-Monocarboxylate transporter 3, Anti-Monocarboxylate transporter 4, Anti-Solute carrier family 16 member 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, human, mouse

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

AEEEKLHKPPADSGVDLREVEHFLKAEPEKNGEVVHTPETSV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC16A3(9123)

General description

The gene SLC16A3 (solute carrier family 16 member 3) is mapped to human chromosome 17q25.3. It belongs to the SLC16A family and is a transmembrane protein. SLC16A3 is expressed in testis, small intestine, lung, brain, heart, kidney, and spleen. SLC16A3 is commonly referred to as MCT4 (monocarboxylate transporter 4).

Immunogen

Monocarboxylate transporter 4 recombinant protein epitope signature tag (PrEST)

Application

Anti-SLC16A3 antibody produced in rabbit has been used in: immunohistochemistry, flow cytometry, and immunofluorescence (IF) confocal microscopy to stain human bronchial epithelial cells (HBECs)-short hairpin RNA (shRNA) targeting TP53(shp53 (shp53)-V5-TMPRSS11B cells.

Biochem/physiol Actions

MCTs (monocarboxylate transporters) are important for transporting lactate and other monocarboxylates across the cell membrane. MCT4/SLC16A3 (solute carrier family 16 member 3) is involved in the transport of γ-hydroxybutyric acid and L-lactate. SLC16A3 is up-regulated in triple negative breast cancer. It is responsible for maintenance of pH, lactate secretion and non-oxidative metabolism of glucose in cancer cells. In similar manner, androgens induce glycolytic metabolism and lactate export in prostate cancer cells by regulating SLC16A3 expression.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75671

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Doyen et al.
Biochemical and biophysical research communications, 451(1), 54-61 (2014-07-25)
(18)Fluor-deoxy-glucose PET-scanning of glycolytic metabolism is being used for staging in many tumors however its impact on prognosis has never been studied in breast cancer. Glycolytic and hypoxic markers: glucose transporter (GLUT1), carbonic anhydrase IX (CAIX), monocarboxylate transporter 1 and
Chantale Farah et al.
Biomedicines, 10(3) (2022-03-26)
A vast majority of BRAF V600E mutated melanoma patients will develop resistance to combined BRAF/MEK inhibition after initial clinical response. Resistance to targeted therapy is described to be accompanied by specific metabolic changes in melanoma. The aim of this work
Cátia V Vaz et al.
Journal of cancer research and clinical oncology, 142(1), 5-16 (2015-06-07)
The present study aims to investigate the role of androgens in controlling the glycolytic metabolism and lactate efflux in prostate cancer (PCa) cells. Androgen-responsive LNCaP cells were treated with 5α-dihydrotestosterone (DHT, 10 nM) for 12-48 h, and their glycolytic metabolism, lactate production
Chantale Farah et al.
International journal of molecular sciences, 24(3) (2023-02-12)
There is currently no consensus to determine which advanced melanoma patients will benefit from immunotherapy, highlighting the critical need to identify early-response biomarkers to immune checkpoint inhibitors. The aim of this work was to evaluate in vivo metabolic spectroscopy using
Ying-Hui Tong et al.
Translational cancer research, 10(3), 1336-1345 (2022-02-05)
The monocarboxylate transporter (MCT) family especially MCT1 and MCT4 have been recognized to play an important role in lactate transport, a key glycolytic product. The expression of MCT1 and MCT4 expression was previously found to be related to poor outcome

Articles

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. MA
    • Reviews 2
    • Votes 0
    5 out of 5 stars.

    Lab

    Frozen section human tissue with 1:800 IHC result look clear and positive signal very specific

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service