Skip to Content
Merck
All Photos(7)

Key Documents

HPA021149

Sigma-Aldrich

Anti-MRPS30 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-28S ribosomal protein S30, mitochondrial, Anti-MRP-S30, Anti-Programmed cell death protein 9, Anti-S30mt

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

FAWTGAQAMYQGFWSEADVTRPFVSQAVITDGKYFSFFCYQLNTLALTTQADQNNPRKNICWGTQSKPLYETIEDNDVKGFNDDVLLQIVHFLLNRP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MRPS30(10884)

General description

The gene MRPS30 (28S ribosomal protein S30, mitochondrial) is mapped to human chromosome 5q11. MRPS30 transcript is ubiquitously expressed with stronger expression in heart, skeletal muscle, kidney, and liver.

Immunogen

28S ribosomal protein S30, mitochondrial recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Mitochondrial ribosomes are composed of small (28S) and large (39S) subunits. MRPS30 (28S ribosomal protein S30, mitochondrial) was initially showed to be component of 28S subunit, however, recent studies showed MRPS30 presence in 39S subunit. Single nucleotide polymorphism is MRPS30 is associated with breast cancer risk.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74175

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Basil J Greber et al.
Nature, 515(7526), 283-286 (2014-10-02)
Mitochondrial ribosomes (mitoribosomes) are extensively modified ribosomes of bacterial descent specialized for the synthesis and insertion of membrane proteins that are critical for energy conversion and ATP production inside mitochondria. Mammalian mitoribosomes, which comprise 39S and 28S subunits, have diverged
E Cavdar Koc et al.
The Journal of biological chemistry, 276(22), 19363-19374 (2001-03-30)
Identification of all the protein components of the small subunit (28 S) of the mammalian mitochondrial ribosome has been achieved by carrying out proteolytic digestions of whole 28 S subunits followed by analysis of the resultant peptides by liquid chromatography
L Carim et al.
Cytogenetics and cell genetics, 87(1-2), 85-88 (2000-01-21)
We report the sequence, tissue distribution, and chromosome location of a novel gene, PDCD9 (programmed cell death 9). PDCD9 is the mammalian counterpart of the Gallus gallus pro-apoptotic protein p52. The human cDNA has an open reading frame of 1,314
Yan-Ping Guan et al.
Asian Pacific journal of cancer prevention : APJCP, 15(1), 85-91 (2014-02-18)
Genome-wide association studies (GWAS) have identified various genetic susceptibility loci for breast cancer based mainly on European-ancestry populations. Differing linkage disequilibrium patterns exist between European and Asian populations. Ten SNPs (rs2075555 in COL1A1, rs12652447 in FBXL17, rs10941679 in 5p12/MRPS30, rs11878583

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service