Skip to Content
Merck
All Photos(7)

Key Documents

HPA019779

Sigma-Aldrich

Anti-GSTP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-GST class-pi, Anti-GSTP1-1, Anti-Glutathione S-transferase P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
clone:
polyclonal
species reactivity:
human
citations:
5
technique(s):
immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

KTFIVGDQISFADYNLLDLLLIHEVLAPGCLDAFPLLSAYVGRLSARPKLKAFLASPEYVNLPINGNGKQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GSTP1(2950)

General description

GSTP1 (Glutathione S-transferase pi 1) is a novel cyclin dependent kinase-5 regulatory protein encoding a detoxifying enzyme. It is widely expressed in normal tissues.

Immunogen

Glutathione S-transferase P recombinant protein epitope signature tag (PrEST)

Application

Anti-GSTP1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

GSTP1 (Glutathione S-transferase pi 1) plays an important role in Cdk signaling pathway by protecting cells from DNA adduct formation. In prevention of neurodegeneration, it blocks activity of Cdk5 via p25/p35 translocation. It has been predicted that GSTP1 level may control Cdk5 signaling, and eliminate oxidative stress. It can also couple with a wide number of exogenous and endogenous hydrophobic electrophiles. Overexpression of enzyme activity has been reported in several types of cancers, such as blood, head and neck, lung, colorectal, esophagus and breast cancers.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74590

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yusuke Yamamoto et al.
Oncology reports, 30(4), 1687-1694 (2013-07-12)
Glutathione S-transferases (GSTs) have been reported to be activated in several types of cancers, including esophageal squamous cell carcinoma (ESCC). The aim of the present study was to investigate whether GSTP1 protein expression is a useful predictor of the clinical
Maja Ludvigsen et al.
Current issues in molecular biology, 43(2), 1043-1056 (2021-09-26)
Colorectal cancer (CRC) is one of the leading causes of cancer-related death over the world. There is a great need for biomarkers capable of early detection and as targets for treatment. Differential protein expression was investigated with two-dimensional gel electrophoresis
Kai-Hui Sun et al.
Journal of neurochemistry, 118(5), 902-914 (2011-06-15)
Cyclin dependent kinase-5 (Cdk5) activity is deregulated in Alzheimer's disease (AD) and contributes to all three hallmarks: neurotoxic β-amyloid formation, neurofibrillary tangles, and neuronal death. However, the mechanism leading to Cdk5 deregulation remains controversial. Cdk5 deregulation in AD is usually
D Pellacani et al.
Cell death and differentiation, 21(5), 761-773 (2014-01-28)
Prostate cancer (CaP) is mostly composed of luminal-like differentiated cells, but contains a small subpopulation of basal cells (including stem-like cells), which can proliferate and differentiate into luminal-like cells. In cancers, CpG island hypermethylation has been associated with gene downregulation
Nicolaas J Bekker et al.
American journal of physiology. Lung cellular and molecular physiology, 324(4), L521-L535 (2023-02-23)
Lung fibroblasts are implicated in abnormal tissue repair in chronic obstructive pulmonary disease (COPD). The exact mechanisms are unknown and comprehensive analysis comparing COPD- and control fibroblasts is lacking. The aim of this study is to gain insight into the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service